1.05289
SLANETZ and BARTLEY agar (base)
acc. ISO 7899 GranuCult
Synonym(s):
Membrane filtration Enterococcus selective agar
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
Recommended Products
Quality Level
form
medium granules (dehydrated (DCM))
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
pH
7.2 (35 °C, 41 g/L in H2O)
solubility
41.5 g/L
bulk density
580 kg/m3
application(s)
food and beverages
microbiology
storage temp.
15-25°C
suitability
Enterococcus spp.
Related Categories
General description
GranuCult is our new brand name for our existing granulated dehydrated culture media which apply to new EN ISO 11133:2014 and various other ISO standards and FDA-BAM / USDA-FSIS methods for testing of food and water samples. All these media are currently being updated to be fully compliant to the latest version of these standards and regulations with respect to composition and quality control and will be available shortly.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service