Skip to Content
Merck
All Photos(1)

Key Documents

1.02700

Supelco

Copper

foil about 0.1 mm thickness for analysis EMSURE®

Synonym(s):

Copper

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
12141711
EC Index Number:
231-159-6
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99.7% (iodometric)

form

solid

impurities

≤0.02% Substances insoluble in nitric acid

bp

2580 °C/1013 hPa

mp

1083 °C

density

8.96 g/cm3

bulk density

1000 kg/m3

cation traces

Ag: ≤0.002%
As: ≤0.0002%
Fe: ≤0.005%
Mn: ≤0.005%
Pb: ≤0.05%
Sb: ≤0.005%
Sn: ≤0.005%

storage temp.

2-30°C

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Application


  • Electron-electron scattering limits thermal conductivity of metals under extremely high electron temperatures: The study discusses how copper′s thermal properties are influenced under extreme conditions, applicable in high-performance electronics (Karna and Giri, 2024).

  • Lateral Flow Assay Biotesting by Utilizing Plasmonic Nanoparticles Made of Inexpensive Metals: Demonstrates copper′s potential as a cost-effective alternative to gold in diagnostic assays, broadening access to healthcare technologies (Bahamondes Lorca et al., 2024).

Analysis Note

Assay (iodometric): ≥ 99.7 %
Substances insoluble in nitric acid: ≤ 0.02 %
Ag (Silver): ≤ 0.002 %
As (Arsenic): ≤ 0.0002 %
Fe (Iron): ≤ 0.005 %
Mn (Manganese): ≤ 0.005 %
Pb (Lead): ≤ 0.05 %
Sb (Antimony): ≤ 0.005 %
Sn (Tin): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service