1.01513
Hydrofluoric acid
48%, Ultrapur
Synonym(s):
Hydrofluoric acid solution, Fluoric acid
About This Item
Recommended Products
product name
Hydrofluoric acid 48%, Ultrapur
grade
for inorganic trace analysis
Quality Level
Assay
48%
form
liquid
pH
2 (20 °C in H2O)
bp
ca.106 °C/ca.223 °F-at 1,013 hPa
mp
-35 °C
solubility
soluble (Water: Soluble)
density
1.16 g/cm3 at 20 °C
cation traces
Ag: ≤20 ppt
Al: ≤100 ppt
As: ≤20 ppt
Au: ≤20 ppt
Ba: ≤20 ppt
Be: ≤20 ppt
Bi: ≤-20 ppt
Ca: ≤500 ppt
Cd: ≤50 ppt
Ce: ≤20 ppt
Co: ≤20 ppt
Cr: ≤100 ppt
Cu: ≤500 ppt
Dy: ≤20 ppt
Er: ≤20 ppt
Eu: ≤20 ppt
Fe: ≤500 ppt
Ga: ≤20 ppt
Gd: ≤20 ppt
Hf: ≤10 ppt
Ho: ≤20 ppt
In: ≤20 ppt
K: ≤200 ppt
La: ≤20 ppt
Li: ≤20 ppt
Lu: ≤20 ppt
Mg: ≤100 ppt
Mn: ≤20 ppt
Mo: ≤20 ppt
Na: ≤200 ppt
Nb: ≤20 ppt
Nd: ≤20 ppt
Ni: ≤50 ppt
Pb: ≤50 ppt
Pd: ≤20 ppt
Pr: ≤20 ppt
Pt: ≤20 ppt
Rb: ≤20 ppt
Re: ≤10 ppt
Sb: ≤20 ppt
Sc: ≤20 ppt
Sm: ≤20 ppt
Sn: ≤50 ppt
Sr: ≤20 ppt
Ta: ≤50 ppt
Tb: ≤20 ppt
Te: ≤50 ppt
Th: ≤1 ppt
Ti: ≤100 ppt
Tl: ≤20 ppt
Tm: ≤20 ppt
U: ≤1 ppt
V: ≤20 ppt
W: ≤100 ppt
Y: ≤10 ppt
Yb: ≤20 ppt
Zn: ≤200 ppt
Zr: ≤20 ppt
SMILES string
F
storage temp.
2-30°C
InChI
InChI=1S/FH/h1H
InChI key
KRHYYFGTRYWZRS-UHFFFAOYSA-N
Application
- Detection of Organic Vapors Based on Photoluminescent Bragg-Reflective Porous Silicon Interferometer: This study demonstrates the use of 48% hydrofluoric acid in fabricating porous silicon structures for sensing organic compounds, showcasing its importance in developing advanced materials for chemical sensing (Ahn et al., 2015).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service