Skip to Content
Merck
All Photos(1)

Key Documents

1.01097

Millipore

Aluminium oxide 90 standardized

for column chromotographic adsorption analysis acc. to Brockmann

Synonym(s):

Aluminium oxide 90 standardized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al2O3
CAS Number:
Molecular Weight:
101.96
MDL number:
UNSPSC Code:
41115711
EC Index Number:
215-691-6

material

aluminum oxide (irregularly-shaped)

Quality Level

description

for column chromotographic adsorption analysis acc. to Brockmann

potency

>5000 mg/kg LD50, oral (Rat)

matrix active group

Irregular-Shaped Aluminum Oxide

particle size

63-200 μm

pore size

9 nm mean pore size

bp

2980 °C/1013 hPa

mp

2050 °C

application(s)

plasma purification

separation technique

normal phase

storage temp.

no temp limit

SMILES string

[Al+3].[Al+3].[O-2].[O-2].[O-2]

InChI

1S/2Al.3O/q2*+3;3*-2

InChI key

PNEYBMLMFCGWSK-UHFFFAOYSA-N

General description

A versatile sorbent for preparative chromatography exhibiting a higher pH stability

Features and Benefits

Aluminium oxide 90 standardized provides users with:

An aluminum oxide crystal structure comprises octahedrally and tetrahedrally coordinated aluminum coupled by oxygen atoms. The surface is covered by free hydroxyl groups. There are different acidic and basic centers that result in anion and cation exchange properties. Aluminum oxide exhibits a higher pH-stability than silica gel, especially in the alkaline range and occurs in various crystal modifications resulting in different pore diameters.

Packaging

Plastic bottle

Analysis Note

pH-value (10 % suspension): 9.0 - 10.0
Substances extractable by water: ≤ 0.5 %
Chloride (Cl): ≤ 0.1 %
Sulfate (SO₄): ≤ 0.1 %
Speed of filtration: ≥ 0.2 ml/min
Activity: II - III
Particle size: 70 % between 0.063 - 0.200 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service