Skip to Content
Merck
All Photos(1)

Key Documents

1.00657

Supelco

Silicic acid

precipitated extra pure light

Synonym(s):

Silicic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si · xH2O
CAS Number:
Molecular Weight:
60.08 (anhydrous basis)
MDL number:
UNSPSC Code:
12352308
EC Index Number:
231-545-4
NACRES:
NB.21

Quality Level

Assay

99-100.5% SiO2 basis (calculated on ignited substance, gravimetric)

form

solid

potency

>10000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

loss

≤8.5% loss on ignition, 2 h, 900 °C

pH

3.0-8.0 (3.3% in water)

mp

1710 °C

solubility

water: insoluble at 20 °C

density

2-6 g/cm3 at 20 °C

bulk density

60‑600 kg/m3

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.5%

cation traces

Fe: ≤0.03%
heavy metals (as Pb): ≤0.0025%

application(s)

sample preparation

storage temp.

no temp limit

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (gravimetric, SiO₂, calc. on ignited substance): 99.0 - 100.5 %
Identity: passes test
pH-value (3.3 %, water): 3.0 - 8.0
Chloride (Cl): ≤ 0.05 %
Sulfate (SO₄): ≤ 0.5 %
Heavy metals (as Pb): ≤ 0.0025 %
Fe (Iron): ≤ 0.03 %
Loss on ignition (2 h, 900 °C): ≤ 8.5 %
Particle size (< 0.1 mm): about 99
Bulk density: about 6

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service