Skip to Content
Merck
All Photos(2)

Documents

09-820

Sigma-Aldrich

Anti-Klf2 Antibody

serum, from rabbit

Synonym(s):

Kruppel-like factor 2 (lung), Lung krueppel-like factor, krueppel-like factor 2, lung Kruppel-like zinc finger transcription factor, Kruppel-like factor LKLF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Klf2(16598)

General description

Krueppel-like factor 2 (Klf2) is a novel protein belonging to the KLF family of transcription factors, a subfamily of approximately 20 members characterized by their possession of a zinc-finger domain and functioning as transcriptional regulators for cellular differentiation and growth. Klf2 activates transcription through its binding of the beta-globin gene promoter CACCC box. Studies have shown Klf2 to have critical roles in both the function and differentiation of several cell types including; adipocytes, lung cells, T lymphocytes, and monocytes. It has also been newly identified as an endothelial transcription regulator for pro-inflammatory activation, and is a requirement for the migration of smooth muscle cells and normal vessel formation. Shear stress and pro-inflammatory cytokines are considered to be potent inhibitors of Klf2. Expression is primarily observed in lung and spleen tissue, but it has also been found in aortic tissue.

Immunogen

Epitope: Unknown
GST-fusion protein with histidine-tagged mouse Klf2.

Application

Chromatin Immunoprecipitation Analysis: 5 µL from a representative lot immunoprecipitated Klf2 from 2 x 10E6 to 1 x 10E7 mouse embryonic stem cells. (Huck Ng, Genome Institute of Singapore.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-Klf2 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect Klf2 also known as Kruppel-like factor 2 (lung), Lung krueppel-like factor.

Quality

Evaluated by Western Blot in mouse embryonic stem cell chromatin extract.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Klf2 in 10 µg of mouse embryonic stem cell chromatin extract.

Target description

~40 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse embryonic stem cell chromatin extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhong Huang et al.
Journal of lipid research, 65(1), 100472-100472 (2023-11-11)
Liver steatosis is a common metabolic disorder resulting from imbalanced lipid metabolism, which involves various processes such as de novo lipogenesis, fatty acid uptake, fatty acid oxidation, and VLDL secretion. In this study, we discovered that KLF2, a transcription factor
Dongbo Qiu et al.
Stem cell reports, 5(3), 314-322 (2015-09-01)
Activation of Wnt/β-catenin signaling can induce both self-renewal and differentiation in naive pluripotent embryonic stem cells (ESCs). To gain insights into the mechanism by which Wnt/β-catenin regulates ESC fate, we screened and characterized its downstream targets. Here, we show that
Yuki Hirata et al.
Biomedical research (Tokyo, Japan), 40(2), 67-78 (2019-04-16)
T1R1 and T1R3 are receptors expressed in taste buds that detect L-amino acids. These receptors are also expressed throughout diverse organ systems, such as the digestive system and muscle tissue, and are thought to function as amino acid sensors. The
Klf2 is an essential factor that sustains ground state pluripotency.
Yeo, JC; Jiang, J; Tan, ZY; Yim, GR; Ng, JH; Goke, J; Kraus, P; Liang, H; Gonzales et al.
Cell Stem Cell null
Chenfeng Liu et al.
Science advances, 7(41), eabg6262-eabg6262 (2021-10-09)
Molecular pathways controlling emigration of mature thymocytes from thymus to the periphery remain incompletely understood. Here, we show that T cell–specific ablation of glycogen synthase kinase 3 (GSK3) led to severely impaired thymic egress. In the absence of GSK3, β-catenin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service