Skip to Content
Merck
All Photos(1)

Key Documents

07-623

Sigma-Aldrich

Anti-Clara Cell Secretory Protein Antibody

serum, Upstate®

Synonym(s):

Anti-CC10, Anti-CCPBP, Anti-CCSP, Anti-UGB, Anti-UP-1, Anti-UP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes Clara Cell Secretory Protein.

Immunogen

A synthetic peptide corresponding to amino acids 45-68 of mouse CCSP coupled to KLH

Application

Detect Clara Cell Secretory Protein using this Anti-Clara Cell Secretory Protein Antibody validated for use in IH & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

routinely evaluated by immunoblot in mouse lung nuclear extracts

Target description

10 kDa

Physical form

Rabbit antiserum containing 0.01% sodium azide and 30% glycerol.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Lung tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lung tissue regeneration after induced injury in Runx3 KO mice.
Jong-Min Lee,Hyuk-Jae Kwon,Suk-Chul Bae,Han-Sung Jung
Cell and Tissue Research null
Loss of Rab27 function results in abnormal lung epithelium structure in mice.
Bolasco, G; Tracey-White, DC; Tolmachova, T; Thorley, AJ; Tetley, TD; Seabra, MC; Hume, AN
American Journal of Physiology. Cell Physiology null
Clinical aspects of Clara cell 10-kDa protein/ uteroglobin (secretoglobin 1A1)
Shijubo, Noriharu, et al
Current Pharmaceutical Design, 9, 1139-1149 (2003)
Local fetal lung renin-angiotensin system as a target to treat congenital diaphragmatic hernia.
Nogueira-Silva, C; Carvalho-Dias, E; Piairo, P; Nunes, S; Baptista, MJ; Moura, RS; Correia-Pinto, J
Molecular Medicine null
Zhenguang Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(5), 6226-6238 (2019-02-23)
Pulmonary airway epithelial cells (AECs) form a critical interface between host and environment. We investigated the role of the circadian clock using mice bearing targeted deletion of the circadian gene brain and muscle ARNT-like 1 (Bmal1) in AECs. Pulmonary neutrophil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service