Skip to Content
Merck
All Photos(3)

Key Documents

07-436

Sigma-Aldrich

Anti-monomethyl-Histone H3 (Lys4) Antibody

Upstate®, from rabbit

Synonym(s):

H3K4me1, Histone H3 (mono methyl K4), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, vertebrates

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys4)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species to cross-reactivity is expected.
Recognizes monomethyl-histone H3 (Lys4), Mr 17kDa.

Immunogen

Epitope: Lys4
KLH-conjugated, synthetic peptide containing the sequence ART[meK]…, in which meK corresponds to monomethyl-lysine at residue 4 of human histone H3.

Application

Anti-monomethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of monomethyl-Histone H3 (Lys4) also known as H3K4me1, Histone H3 (mono methyl K4) & has been validated in ICC, IF, PIA, WB, ChIP-seq, DB.
Immunocytochemistry:
Reported by an independent laboratory.

Peptide Inhibition:
Specificity was confirmed by the ability of 1 μM of the immunizing peptide to completely abolish detection of histone H3 in immunoblot analysis of HeLa acid extracts. (see figure, lane 3). No competition was observed with peptides containing dimethyl-lysine 4, trimethyl-lysine 4, mono-methyllysine 4 or monomethyl-lysine 9 or 27.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by Western Blot in acid extracted proteins from HeLa cells.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected methylated histone H3 in acid extracted proteins from HeLa cells.

Target description

17 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05 Sodium Azide, pH 7.4.

Storage and Stability

Stable for 1 year at 2 to 8°C from date of receipt.

Analysis Note

Control
HeLa cell extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Hsin Su et al.
PloS one, 11(11), e0166090-e0166090 (2016-11-03)
Cytokines are low molecular weight regulatory proteins, or glycoproteins, with both tumor-promoting and inhibitory effects on breast cancer growth. Different cytokines play important roles in breast cancer initiation and progression. Here, we show that of the 39 interleukin (IL) genes
PU.1 and C/EBP(alpha) synergistically program distinct response to NF-kappaB activation through establishing monocyte specific enhancers.
Jin, F; Li, Y; Ren, B; Natarajan, R
Proceedings of the National Academy of Sciences of the USA null
Arabidopsis trithorax-related3/SET domain GROUP2 is required for the winter-annual habit of Arabidopsis thaliana.
Yun, JY; Tamada, Y; Kang, YE; Amasino, RM
Plant & Cell Physiology null
Emily Petty et al.
PloS one, 6(10), e25973-e25973 (2011-10-15)
Dosage compensation is a specialized form of gene regulation that balances sex-chromosome linked gene expression between the sexes. In C. elegans, dosage compensation is achieved by the activity of the dosage compensation complex (DCC). The DCC binds along both X
Jong-Hyun Ko et al.
The EMBO journal, 29(18), 3208-3215 (2010-08-17)
In Arabidopsis, the rapid-flowering summer-annual versus the vernalization-requiring winter-annual growth habit is determined by natural variation in FRIGIDA (FRI) and FLOWERING LOCUS C (FLC). However, the biochemical basis of how FRI confers a winter-annual habit remains elusive. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service