Skip to Content
Merck
All Photos(1)

Documents

07-228

Sigma-Aldrich

Anti-TACC2 Antibody

Upstate®, from rabbit

Synonym(s):

transforming, acidic coiled-coil containing protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TACC2(10579)

General description

TACC2, also known as transforming acidic coiled-coil-containing protein 2, belongs to the transforming acidic coiled-coil-containing (TACC) protein family. This group of proteins is defined by its highly conserved coiled-coil domain. This domain is necessary for the assignment of the centrosome-spindle apparatus during cell cycling. There are three proteins belonging to the TACC family; TACC1, TACC2, and TACC3. TACC2 has been reported as playing a necessary role in cultured cell microtubule stabilization. It has also been shown that the binding of TACC2 by T antigen can cause chromosomal instability and mitotic defects (Tei, 2009). Research into the over expression of TACC2 strongly suggests a potential function as a tumor suppressor (Tei, 2009).

Immunogen

KLH-conjugated linear peptide corresponding to the mature chain of mouse TACC2.

Application

Anti-TACC2 Antibody is a Rabbit Polyclonal Antibody for detection of TACC2 also known as transforming acidic coiled-coil containing protein 2 & has been validated in WB.

Quality

Evaluated by Western Blot in mouse testis tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected TACC2 on 10 µg of mouse testis tissue lysate.

Target description

Observed molecular weight, 73 kDa
Note this protein can be highly modified and several isoforms exist.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Wong et al.
Journal of applied microbiology, 103(4), 803-810 (2007-09-28)
To survey the prevalence of Salmonella in imported and domestic pet chews for assessing their potential in introducing novel pathogenic and antimicrobial resistant Salmonella serotype clones into New Zealand, and as vehicles of salmonellosis in the domestic home environment. Three
T L Buhr et al.
Journal of applied microbiology, 115(2), 398-408 (2013-05-23)
To develop test methods and evaluate survival of Bacillus anthracis Ames, B. anthracis ∆Sterne and B. thuringiensis Al Hakam spores after exposure to PES-Solid (a solid source of peracetic acid), including PES-Solid formulations with bacteriostatic surfactants. Spores (≥ 7 logs)
M Myrmel et al.
Journal of fish diseases, 37(4), 349-355 (2013-04-27)
The aquaculture industry needs a simple, inexpensive and safe method for the treatment of fish waste without heat. Microbial inactivation by inorganic acid (HCl) or base (KOH) was determined using infectious pancreatic necrosis virus (IPNV) as a model organism for
Ken-ichi Takayama et al.
Molecular endocrinology (Baltimore, Md.), 26(5), 748-761 (2012-03-30)
Despite the existence of effective antiandrogen therapy for prostate cancer, the disease often progresses to castration-resistant states. Elucidation of the molecular mechanisms underlying the resistance for androgen deprivation in terms of the androgen receptor (AR)-regulated pathways is a requisite to
Molecular cloning, genomic structure and interactions of the putative breast tumor suppressor TACC2.
Brenda Lauffart, Omkaram Gangisetty, Ivan H Still
Genomics null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service