Skip to Content
Merck
All Photos(2)

Key Documents

06-578-I

Sigma-Aldrich

Anti-PI4-Kinase β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Unknown, polyclonal

mol wt

calculated mol wt 91.38 kDa
observed mol wt ~92 kDa

purified by

affinity chromatography

species reactivity

human, monkey

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

rabbit ... PI4KB(5298)

General description

Phosphatidylinositol 4-kinase beta (UniProt: Q9UBF8; also known as EC:2.7.1.67, PI4K-beta, PI4Kbeta, PtdIns 4-kinase beta, NPIK, PI4K92, PI4KIII) is encoded by the PI4KB (also known as PIK4CB) gene (Gene ID: 5298) in human. A group of four enzymes make up the mammalian phosphatidylinositol (PI) 4-kinase family are divided into two groups based on primary sequence and biochemical properties: consisting of type II (PI4KII and PI4KIIβ) and the type III (PI4KIII and PI4KIIIβ) isozymes. They all catalyze the phosphorylation of phosphatidylinositol (PI) at the D4 position of the inositol head-group to synthesize phosphatidylinositol 4-phosphate (PI4P) that serves as an essential precursor to produce PIP2 and PIP3. PI4-kinase b is a type III peripheral membrane protein that is present in the outer membrane of mitochondria and membranes of the rough endoplasmic reticulum. It phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1,4,5,-trisphosphate (PIP). It also regulates Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation and is involved in Golgi-to-plasma membrane trafficking. PI4-kinase b is shown to be critical for glucose-induced insulin secretion due to its capacity to regulate the release of secretory granules. In addition, it inhibits the insulin-stimulated translocation of glucose transporter-4 in adipocytes through its interaction with neuronal calcium sensor. (Ref.: Chalupska, D., et al. (2019). Sci. Rep. 9; article 567; Clyton, EL., et al. (2013). Prog. Lipid Res. 52(3); 294-304).

Specificity

This rabbit polyclonal antibody specifically detects Phosphatidylinositol 4-kinase b (PI4-kinase b). It targets an epitope within 18 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal region of human Phosphatidylinositol 4-kinase b (PI4-Kinase b).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PI4-Kinase β in Jurkat cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PI4-Kinase β in HeLa and SH-SY5Y cell lysates.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected PI4-Kinase β in THP-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-PI4-Kinase β , Cat. No. 06-578-I, is a rabbit polyclonal antibody that detects Phosphatidylinositol 4-kinase b and is tested for use in Immunocytochemistry and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service