Skip to Content
Merck
All Photos(1)

Key Documents

06-207

Sigma-Aldrich

Anti-Lyn Antibody

serum, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LYN(4067)

Specificity

two known forms of human Lyn

Immunogen

peptide (NKQQRPVPESQL-LPGQRFQT) corresponding to residues 39-58 of human Lyn

Application

Anti-Lyn Antibody detects level of Lyn & has been published & validated for use in IP & WB.

Quality

routinely evaluated by immunoblot on RIPA lysates from human A431 cells

Target description

53 and 56kDa

Physical form

whole antiserum containing 0.05% sodium azide

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Vonakis et al.
The Journal of biological chemistry, 272(38), 24072-24080 (1997-09-20)
Aggregation of the high affinity receptor for IgE (FcepsilonRI) leads to the phosphorylation of tyrosines on the beta and gamma chains of the receptor by the Src family kinase Lyn. We have studied the interaction between Lyn and the FcepsilonRI
Mirela Goldenberg-Furmanov et al.
Cancer research, 64(3), 1058-1066 (2004-02-12)
The Src-related protein kinase Lyn plays an important role in B-cell activation. However, several lines of evidence suggest that it is also involved in the control of cellular proliferation and the inhibition of apoptosis. We have discovered that Lyn is
P Kumar et al.
The Journal of biological chemistry, 276(24), 21039-21045 (2001-03-29)
Cellular adhesion molecules such as E-selectin function to recruit leukocytes into the inflammatory lesions of diseases such as rheumatoid arthritis (RA) and atherosclerosis. Monocytes are the key components of the cellular infiltrates present in these disorders. We hypothesized that soluble
J Holland et al.
The Journal of biological chemistry, 272(14), 9108-9112 (1997-04-04)
Intercellular adhesion molecule 1 (ICAM-1) (CD54) is an adhesion molecule of the immunoglobulin superfamily. The interaction between ICAM-1 on B lymphocytes and leukocyte function-associated antigen 1 on T cells plays a major role in several aspects of the immune response
Genetic evidence for a tyrosine kinase cascade preceding the mitogen-activated protein kinase cascade in vertebrate G protein signaling
Wan, Y., et al
The Journal of Biological Chemistry, 272, 17209-17215 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service