Skip to Content
Merck
All Photos(4)

Key Documents

06-1106

Sigma-Aldrich

Anti-Sonic Hedgehog Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

sonic hedgehog, sonic hedgehog (Drosophila) homolog, sonic hedgehog homolog (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SHH(6469)

General description

Sonic Hedgehog Protein (SHH, VHH-1) belongs to hedgehog protein family which includes Indian Hh, and Desert Hh. Hh family is involved in the cell fate and patterning during embryonic development, homeostasis, and adult tissue renewal. Similar to other member in the family, SHH binds to the patched (PTC) cell surface receptor, releasing the signal transducer Smoothened (Smo) to transmit the Hh signal into the cell and activate transcription of the target gene. Precursor SHH is autocatlytically cleaved into two subunits, N-terminal and C-terminal products. Soluble N-terminal product is involved in the signaling activity, while C-product displays an autoproteolysis activity on the precursor and a cholesterol transferase activity on the N-terminal product. C-terminal product attaches a cholesterol moiety to the N-terminal product, preventing N-terminal product diffusion within the developing embryo. A defect in SHH has been linked in holoprosencephaly type 3 (HPE3), in which developing forebrain fails to separate into right and left hemispheres, and ocular coloboma.

Specificity

This antibody recognizes the N-terminus of Sonic Hedgehog.

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to the N-terminus of Sonic HedgeHog.

Application

Anti-Sonic Hedgehog Antibody detects level of Sonic Hedgehog & has been published & validated for use in WB, IH.
Research Category
Signaling
Research Sub Category
Developmental Signaling
Western Blot (SNAP ID) Analysis: 1 µg/mL from a previous lot detected Sonic Hedgehog on 10 µg of human fetal skeletal muscle tissue lysate.

Immunohistochemistry Analysis: 1:500 dilution from a previous lot detected Sonic Hedgehog in colorectal carcinoma tissue.

Quality

Evaluated by Western Blot in human fetal skeletal muscle tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected Sonic Hedgehog on 10 µg of human fetal skeletal muscle tissue lysate.

Target description

~ 45 kDa

Linkage

Replaces: 04-971

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human fetal skeletal muscle tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of targets for rational pharmacological therapy in childhood craniopharyngioma.
Gump, JM; Donson, AM; Birks, DK; Amani, VM; Rao, KK; Griesinger et al.
Acta Neuropathologica Communications null
Gregory M Shackleford et al.
PloS one, 11(6), e0156907-e0156907 (2016-06-17)
Medulloblastomas are the most common malignant pediatric brain tumor and have been divided into four major molecular subgroups. Animal models that mimic the principal molecular aberrations of these subgroups will be important tools for preclinical studies and allow greater understanding
Chengyun Xu et al.
Mucosal immunology, 11(5), 1306-1315 (2018-06-06)
Sonic hedgehog (SHH) is abundantly expressed and critical for morphogenesis in embryonic lungs; however, SHH expression drops to a much lower level in mice from E17.5 and in humans from the 21st gestational week. We find that SHH expression is
A Almazán-Moga et al.
British journal of cancer, 117(9), 1314-1325 (2017-09-08)
Rhabdomyosarcoma (RMS) is the most common type of soft tissue sarcoma in children. The Hedgehog (HH) pathway is known to develop an oncogenic role in RMS. However, the molecular mechanism that drives activation of the pathway in RMS is not
Ziyue Li et al.
Molecular medicine reports, 15(1), 297-304 (2016-12-14)
Amelogenesis, beginning with thickened epithelial aggregation and ending with highly mineralized enamel formation, is a process mediated by a complex signaling network that involves several molecules, including growth and transcription factors. During early tooth development, the transcription factor B‑cell CLL/lymphoma 11B (Bcl11b)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service