05-629-IM
Anti-Histone H1o/H5 Antibody, clone 3H9
ascites fluid, clone 3H9, from mouse
Synonym(s):
Histone H5, Histone H1o
Select a Size
Select a Size
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
3H9, monoclonal
species reactivity
human, chicken
packaging
antibody small pack of 25 μL
technique(s)
ChIP: suitable (ChIP-chip)
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
Related Categories
1 of 4
This Item | HPA040140 | SAB4500943 | HPA031100 |
---|---|---|---|
affinity isolated antibody | affinity isolated antibody | affinity isolated antibody | affinity isolated antibody |
100 | 100 | 100 | 100 |
rabbit | rabbit | rabbit | rabbit |
Prestige Antibodies® Powered by Atlas Antibodies | Prestige Antibodies® Powered by Atlas Antibodies | - | Prestige Antibodies® Powered by Atlas Antibodies |
polyclonal | polyclonal | polyclonal | polyclonal |
General description
Specificity
Immunogen
Application
Chromatin Immunoprecipitation Analysis: A representative lot detected Histone H1o/H5 in Chromatin Immunoprecipitation applications (Torres, C.M., et. al. (2016). Science. 353(6307)).
Flow Cytometry Analysis: A representative lot detected Histone H1o/H5 in Flow Cytometry applications (Torres, C.M., et. al. (2016). Science. 353(6307)).
Immunocytochemistry Analysis: A representative lot detected Histone H1o/H5 in Immunocytochemistry applications (Torres, C.M., et. al. (2016). Science. 353(6307)).
Immunofluorescence Analysis: A representative lot detected Histone H1o/H5 in Immunofluorescence applications (Torres, C.M., et. al. (2016). Science. 353(6307)).
Quality
Western Blotting Analysis: A 1:20,000 dilution of this antibody detected Histone H1o in 10 µg of HeLa cells acid extract.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service