Skip to Content
Merck
All Photos(1)

Key Documents

05-429

Sigma-Aldrich

Anti-phospho-MBP Antibody, clone P12

clone P12, Upstate®, from mouse

Synonym(s):

Anti-C76307, Anti-R75289, Anti-golli-mbp, Anti-jve, Anti-mld, Anti-shi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P12, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr98)

Gene Information

human ... MBP(4155)

General description

Myelin basic protein (MBP) is the major extrinsic membrane protein of central nervous system and has a molecular mass of 21.5 kDa. MBP is believed to be important in the process of myelination of nerves in the central nervous system (CNS). MBP was initially sequenced in 1979 after isolation from myelin membranes. Since that time, knockout mice deficient in MBP have been developed which showed decreased amounts of CNS myelination and a progressive disorder characterized by tremors, seizures, and early death. The gene for MBP is on chromosome 18; the protein localizes to the CNS and to various cells of the hematopoietic system.
The pool of MBP in the central nervous system is very diverse, with several splice variants being expressed and a large number of post-translational modifications on the protein, which include phosphorylation, methylation, deamidation and citrullination. MBP phosphorylation at Threonine 125 is a complex regulatory process that modulates the contribution of MBP to the stability of the Myelin sheath.
Interest in MBP has centered on its role in demyelinating diseases, particularly multiple sclerosis (MS). Several studies have shown a role for antibodies against MBP in the pathogenesis of MS. Some studies have linked a genetic predisposition to MS to the MBP gene, though a majority have not.

Specificity

Recognizes MBP phosphorylated at Threonine 98; Some preparations of MBP contain basal levels of phosphorylated MBP, which are detected by the pMBP antibody.

Immunogen

Synthetic peptide corresponding to the human myelin basic protein sequence phosphorylated at Thr98 and coupled to tuberculin

Application

Detect phospho-MBP using this Anti-phospho-MBP Antibody, clone P12 validated for use in EA & WB.
Research Category
Signaling
Research Sub Category
MAP Kinases

Quality

routinely evaluated by immunoblot on MBP phosphorylated by MAP Kinase 2/Erk2, active (Catalog #14-173)

Target description

33 kDa

Physical form

Format: Purified
Protein A purified
Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, PBS, and 0.09% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Mouse brain extract or rat brain extract

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cefan Zhou et al.
Cell communication and signaling : CCS, 17(1), 105-105 (2019-08-24)
Breast cancer is a life-threatening disease in females and the leading cause of mortality among the female population, presenting huge challenges for prognosis and treatment. ITM2A is a member of the BRICHOS superfamily, which are thought to have a chaperone
Identification of a mitogen-activated protein kinase site in human myelin basic protein in situ.
Yon, M, et al.
Journal of Neuroimmunology, 65, 55-59 (1996)
Lingfeng Liu et al.
Genes & cancer, 3(1), 23-36 (2012-08-16)
Regulation of Myc protein abundance is critical for normal cell growth as evidenced by the fact that deregulated Myc expression is a hallmark of many cancers. One of several important mechanisms that control Myc levels involves its phosphorylation-dependent proteolysis. Previous
The receptor for advanced glycation end-products (RAGE) directly binds to ERK by a D-domain-like docking site.
Katsuya Ishihara, Kae Tsutsumi, Shiho Kawane, Motowo Nakajima, Tatsuhiko Kasaoka
Febs Letters null
Romina Giacometti et al.
Pest management science, 72(8), 1585-1594 (2015-11-26)
Southern green stink bugs (Nezara viridula L.) invade field-grown soybean crops, where they feed on developing seeds and inject phytotoxic saliva, which causes yield reduction. Although leaf responses to herbivory are well studied, no information is available about the regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service