Skip to Content
Merck
All Photos(1)

Key Documents

05-361

Sigma-Aldrich

Anti-NFκB p52 Antibody

Upstate®, from mouse

Synonym(s):

DNA-binding factor KBF2, Lymphocyte translocation chromosome 10, Oncogene Lyt-10, nuclear factor of kappa light chain gene enhancer in B-cells 2, nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, nuclear factor of kappa light polypept

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

electrophoretic mobility shift assay: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

The NFκB transcription factor was originally identified as a protein complex consisting of a DNA binding subunit and an associated protein. The DNA binding subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino terminus of a precursor designated p105. A cDNA has been isolated that encodes an alternative DNA binding subunit of NFκB. It is synthesized as a protein that is expressed in a variety of cell types and, like p105, undergoes cleavage to generate its NFκB subunit, in this case a protein designated p52 (previously referred to as p49). In contrast to p50 derived from p105, p52 acts in synergy with p65 to stimulate the HIV enhancer in transiently transfected Jurkat cells.

Specificity

Does not cross-react with mouse or rat; other species cross-reactivity is unknown.
This antibody recognizes the p52 subunit of NFκB and its p100 precursor.

Immunogen

GST-fusion protein corresponding to amino acids 1-444 of human NFκB p52 subunit (Schmidt, R., 1991;Bours, V., 1992).

Application

Anti-NFκB p52 Antibody is a Mouse Monoclonal Antibody for detection of NFκB p52 also known as DNA-binding factor KBF2, Lymphocyte translocation chromosome 10, Oncogene Lyt-10 & has been validated in EMSA, IP & WB.
Immunoprecipitation:
4 µg of a previous lot immunoprecipitated NFκB p52 and its p100 precursor from 500 µg of HeLa nuclear extract cell lysate.

Gel Shift Assay:
A previous lot of this antibody used 2 µg per gel shift assay (Bours, V., 1993).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by immunoblotting in HeLa nuclear extract or in human Raji cell, but not in mouse 3T3/A31 cell and rat PC 12 cell lysates

Western Blot Analysis:
0.5-2 µg/mL of this lot detected NFκB p52 and its p100 precursor in HeLa nuclear extract. A previous lot detected NFκB p52 and its p100 precursor in human Raji cell, but not in mouse 3T3/A31 cell and rat PC 12 cell lysates. Highly recommended for western blotting.

Target description

52 kDa; precursor at 100 kDa

Physical form

Format: Purified
Protein G Chromatography
Purified mouse monoclonal IgG2a in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Pulvino et al.
Blood, 120(8), 1668-1677 (2012-07-14)
Diffuse large B-cell lymphoma (DLBCL), the most common type of non-Hodgkin lymphoma, remains a partially curable disease. Genetic alterations affecting components of NF-κB signaling pathways occur frequently in DLBCL. Almost all activated B cell-like (ABC) DLBCL, which is the least
TNF Receptor-Associated Factor 1 is a Major Target of Soluble TWEAK.
Carmona Arana, JA; Seher, A; Neumann, M; Lang, I; Siegmund, D; Wajant, H
Frontiers in Immunology null
Tumor-derived p53 mutants induce NF-kappaB2 gene expression.
Scian, MJ; Stagliano, KE; Anderson, MA; Hassan, S; Bowman, M; Miles, MF; Deb, SP; Deb, S
Molecular and cellular biology null
CD28 delivers a unique signal leading to the selective recruitment of RelA and p52 NF-kappaB subunits on IL-8 and Bcl-xL gene promoters.
Marinari, B; Costanzo, A; Marzano, V; Piccolella, E; Tuosto, L
Proceedings of the National Academy of Sciences of the USA null
Alternative splicing variants of IkappaB beta establish differential NF-kappaB signal responsiveness in human cells
Hirano, F., et al
Molecular and cellular biology, 18, 2596-2607 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service