Skip to Content
Merck
All Photos(1)

Key Documents

04-642

Sigma-Aldrich

Anti-Ago2 Antibody, clone 9E8.2

ascites fluid, clone 9E8.2, Upstate®

Synonym(s):

Argonaute-2, Eukaryotic translation initiation factor 2C, AGO2, eIF-2C, Slicer protein, MGC3183, Piwi domain protein, PPD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
9E8.2, monoclonal
application:
ChIP
WB
species reactivity:
human
technique(s):
ChIP: suitable (ChIP-seq)
western blot: suitable
citations:
23

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

9E8.2, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AGO2(27161)

General description

Ago2 (argonaute-2), also known as eukaryotic translation initiation factor 2C (EIF2C2), is an essential component for siRNA-directed RNA interference (RNAi) response. Ago2 is an endonuclease required for the unwinding of siRNA duplex and assembly of siRNA into RNA-induced silencing complexes (RISC). Ago2 interacts with DICER1 through its Piwi domain. This Piwi domain is thought to provide RNA cleavage activity via a mechanism similar to RNase H. Ago2 activity is necessary for embryonic development as well as RNA-mediated gene silencing (RNAi).

Specificity

human Ago2

Immunogen

Epitope: near the N-terminus of Ago2
KLH-conjugated, synthetic peptide containing the sequence YSGAGPALAPPAPPPPIQG

Application

Anti-Ago2 Antibody, clone 9E8.2 is a Mouse Monoclonal Antibody for detection of Ago2 also known as Argonaute-2, Eukaryotic translation initiation factor 2C & has been validated in WB and has been demonstrated to perform in ChIP and ChIP-seq.
ChIP and ChIP-seq: A representative lot of this antibody was used to perform ChIP and ChIP-seq ( Woolnough, JL, Atwood, BL, and Giles KE (2015), MCB Vol. 35 No. 13, p. 2278-2294).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Chromatin Biology

RNA Binding Protein (RBP)

Quality

Routinely evaluated by immunoblot.

Target description

~100 kda

Physical form

Ascites with 0.05% sodium azide
ImmunoAffinity Purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keriayn N Smith et al.
Stem cell reports, 9(1), 108-121 (2017-06-06)
Of the thousands of long noncoding RNAs expressed in embryonic stem cells (ESCs), few have known roles and fewer have been functionally implicated in the regulation of self-renewal and pluripotency, or the reprogramming of somatic cells to the pluripotent state.
Li Zhang et al.
Cell death & disease, 10(3), 168-168 (2019-02-20)
Cholestasis induces the hepatic long non-coding RNA H19, which promotes the progression of cholestatic liver fibrosis. However, microRNAs that are dysregulated by H19 during cholestasis remain elusive. Using miRNA-sequencing analysis followed by qPCR validation, we identified marked upregulation of eight
Simon J Allison et al.
Molecular therapy. Nucleic acids, 2, e141-e141 (2014-01-09)
Selective gene silencing by RNA interference (RNAi) involves double-stranded small interfering RNA (ds siRNA) composed of single-stranded (ss) guide and passenger RNAs. siRNA is recognized and processed by Ago2 and C3PO, endonucleases of the RNA-induced silencing complex (RISC). RISC cleaves
Differential expression of microRNA expression in tamoxifen-sensitive MCF-7 versus tamoxifen-resistant LY2 human breast cancer cells.
Manavalan, TT; Teng, Y; Appana, SN; Datta, S; Kalbfleisch, TS; Li, Y; Klinge, CM
Cancer letters null
Hanane Ennajdaoui et al.
Cell reports, 15(9), 1876-1883 (2016-05-24)
Insulin-like growth factor 2 mRNA binding protein 3 (IGF2BP3) expression correlates with malignancy, but its role(s) in pathogenesis remains enigmatic. We interrogated the IGF2BP3-RNA interaction network in pancreatic ductal adenocarcinoma (PDAC) cells. Using a combination of genome-wide approaches, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service