Skip to Content
Merck
All Photos(1)

Key Documents

04-119

Sigma-Aldrich

Anti-acetyl-Histone H4 (Lys12) Antibody, rabbit monoclonal

culture supernatant, from rabbit

Synonym(s):

H4K12Ac, Histone H4 (acetyl K12)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, vertebrates

manufacturer/tradename

Chemicon®
Upstate®

technique(s)

ChIP: suitable
dot blot: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys12)

Gene Information

human ... H4C1(8359)

Specificity

Recognizes Histone H4 when acetylated on Lys12.
Wide range of cross-reactivity expected based on sequence homology.

Immunogen

Peptide corresponding to Histone H4 containing the sequence [GLG-AcK-GGA] on which Lys12 is acetylated.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 2 µL of Negative Control Supernatant, or 2 µL of Anti-Acetyl-Histone H4 (Lys12) and the Magna ChIP A Kit (Cat. # 17-610).
Successful immunoprecipitation of Acetyl-Histone H4 (Lys12)-associated DNA fragments was verified by qPCR using ChIP Primers, human GAPDH Coding Region as a positive locus, and a gene desert region as a negative locus. (Figure 2). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Lysates from HeLa cells untreated or sodium butyrate treated (Lanes 1 and 2 respectively) were resolved probed with anti-acetyl-Histone H4 (Lys12) (1:1,000). Arrow indicates Acetyl-Histone H4 (Lys12).
Arrow indicates Acetyl-Histone H4 (Lys12) (~11 kDa)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology
This Anti-acetyl-Histone H4 (Lys12) Antibody, rabbit is validated for use in WB for the detection of acetyl-Histone H4 (Lys12).

Quality

Routinely evaluated by immunoblot.

Target description

11kDa

Physical form

100 μL of rabbit monoclonal IgG cell culture supernatant with 0.1% sodium azide.

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerome Jeanblanc et al.
The international journal of neuropsychopharmacology, 18(9) (2015-03-13)
New strategies for the treatment of alcohol dependence are a pressing need, and recent evidence suggests that targeting enzymes involved in epigenetic mechanisms seems to have great potential. Among these mechanisms, alteration of histone acetylation by histone deacetylases is of
Fasting and high-fat diet alter histone deacetylase expression in the medial hypothalamus.
Funato, H; Oda, S; Yokofujita, J; Igarashi, H; Kuroda, M
Testing null
Changning Wang et al.
Journal of medicinal chemistry, 57(19), 7999-8009 (2014-09-10)
Epigenetic enzymes are now targeted to treat the underlying gene expression dysregulation that contribute to disease pathogenesis. Histone deacetylases (HDACs) have shown broad potential in treatments against cancer and emerging data supports their targeting in the context of cardiovascular disease
Andrew J Kennedy et al.
Cell reports, 16(10), 2666-2685 (2016-08-30)
Human haploinsufficiency of the transcription factor Tcf4 leads to a rare autism spectrum disorder called Pitt-Hopkins syndrome (PTHS), which is associated with severe language impairment and development delay. Here, we demonstrate that Tcf4 haploinsufficient mice have deficits in social interaction
Daniel M Fass et al.
Neuropharmacology, 64, 81-96 (2012-07-10)
Long-term memory formation is known to be critically dependent upon de novo gene expression in the brain. As a consequence, pharmacological enhancement of the transcriptional processes mediating long-term memory formation provides a potential therapeutic strategy for cognitive disorders involving aberrant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service