Skip to Content
Merck
All Photos(1)

Documents

04-078

Sigma-Aldrich

Anti-phospho-Androgen Receptor (Ser81) Antibody, rabbit monoclonal

culture supernatant, Upstate®

Synonym(s):

Anti-AIS, Anti-AR8, Anti-DHTR, Anti-HUMARA, Anti-HYSP1, Anti-KD, Anti-NR3C4, Anti-SBMA, Anti-SMAX1, Anti-TFM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer81)

Gene Information

human ... AR(367)

Specificity

Androgen Receptor when phosphorylated on Ser81.

Immunogen

A synthetic phospho-peptide surrounding Ser81 of human Androgen Receptor.

Application

Anti-phospho-Androgen Receptor (Ser81) Antibody detects level of phospho-Androgen Receptor (Ser81) & has been published & validated for use in WB.

Quality

Routinely evaluated by immunoblot.

Target description

99 kDa

Physical form

100μl of cell culture supernatant with 0.1% sodium azide

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne N Davis et al.
Cancer research, 66(24), 11897-11906 (2006-12-21)
Activation of E2F transcription factors, through disruption of the retinoblastoma (Rb) tumor-suppressor gene, is a key event in the development of many human cancers. Previously, we showed that homozygous deletion of Rb in a prostate tissue recombination model exhibits increased
Shinichiro Yamamoto et al.
Cancer science, 110(11), 3476-3485 (2019-08-28)
Octamer transcription factor 1 (OCT1) is an androgen receptor (AR)-interacting partner and regulates the expression of target genes in prostate cancer cells. However, the function of OCT1 in castration-resistant prostate cancer (CRPC) is not fully understood. In the present study
Shen Gao et al.
Journal of molecular endocrinology, 36(3), 463-483 (2006-05-25)
Androgen receptor (AR) is a ligand-activated transcription factor that mediates the action of androgens and is essential for the growth, function, and cell differentiation of the prostate gland. Here, we demonstrated that the prostate apoptosis response factor-4 (par-4) functions as
Remi Adelaiye-Ogala et al.
Cancer research, 78(11), 2886-2896 (2018-03-25)
Androgen receptor (AR) plays a crucial role in the development and progression of prostate cancer. AR expression has also been reported in other solid tumors, including renal cell carcinoma (RCC), but its biological role here remains unclear. Through integrative analysis
Theodoros Karantanos et al.
Oncotarget, 7(29), 46321-46334 (2016-06-23)
Caveolin-1 (Cav-1) is overexpressed in aggressive and metastatic prostate cancer (PCa) and induces PCa cell proliferation. Androgens mediate lipid synthesis through acetyl-CoA carboxylase-1 (ACC1) and fatty acid synthase (FASN). We investigated the Cav-1-mediated lipid synthesis in the development of castration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service