Skip to Content
Merck
All Photos(1)

Key Documents

T-077

Supelco

3,3′,5-Triiodo-L-thyronine-13C6 solution

100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C9H12I3NO4
CAS Number:
Molecular Weight:
656.93
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 0.1N NH3

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

IC1=CC(O[13C]2=[13C](I)[13CH]=[13C](C[C@H](N)C(O)=O)[13CH]=[13C]2I)=CC=C1O

InChI

1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1/i3+1,4+1,7+1,10+1,11+1,14+1

InChI key

AUYYCJSJGJYCDS-HRUXDFQWSA-N

General description

A Certified Spiking Solution® of the internal standard 3,3´,5-Triiodo-L-thyronine-13C6 (T3-13C6). 3,3′,5-Triiodo-L-thyronine or T3 is a thyroid hormone involved with many physiological processes in the human body including growth and development, metabolism, body temperature and heart rate. This internal standard is suitable for quantitation of free and total T3 serum levels by LC-MS/MS for determination of hyperthyroidism caused by excess T3.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Stefanaki et al.
The Journal of dermatological treatment, 26(2), 139-142 (2014-05-20)
All available treatments for anogenital warts have substantial failure rates. An immunomodulating treatment that enhances the patient's own immunity could be valuable as an adjuvant to conventional methods. About 123 patients were enrolled in this study and were randomized either
Hong Xia et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 23(8), 1648-1655 (2014-05-20)
Although direct transoral decompression and one-stage posterior instrumentation can obtain satisfactory cord decompression for the treatment of basilar invagination with atlantoaxial dislocation, surgical injuries run high as combinative anterior-posterior approaches were necessary. Furthermore, the complications will rise notably when involvement
Ali Tabatabaei et al.
Journal of cataract and refractive surgery, 40(7), 1092-1096 (2014-05-20)
To compare the accuracy of 3 imaging modalities for preoperative evaluation of the posterior lens capsule in traumatic cataract. Farabi Eye Hospital, Tehran, Iran. Case series. The study comprised eyes with traumatic cataract opaque enough to prevent visualization of the
Mitchell S Cappell et al.
Digestive diseases and sciences, 59(10), 2381-2389 (2014-05-20)
To quantitatively describe the syndrome of Mallory-Weiss tears associated (MWa) with antecedent transesophageal echocardiography (TEE) as a distinct syndrome. Cases of MWa were identified by comprehensive, computerized literature search via PubMed and review of textbooks and monographs on TEE and
G Janoray et al.
Cancer radiotherapie : journal de la Societe francaise de radiotherapie oncologique, 18(3), 191-197 (2014-05-20)
Robotic stereotactic body radiation therapy is a new option to treated unresecable liver tumours. The objectives were to assess the tolerance of this technique, to identify predictive factors for toxicity and evaluate the efficiency of this treatment. From June 2010

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service