Skip to Content
Merck
All Photos(1)

Key Documents

T-075

Supelco

3,3′,5′-Triiodo-L-thyronine (Reverse T3) solution

100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

3,3′,5′-Triiodo-L-thyronine, Reverse T3, rT(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 0.1N NH3

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N[C@@H](Cc1ccc(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H12I3NO4/c16-9-3-7(4-12(19)15(21)22)1-2-13(9)23-8-5-10(17)14(20)11(18)6-8/h1-3,5-6,12,20H,4,19H2,(H,21,22)/t12-/m0/s1

InChI key

HZCBWYNLGPIQRK-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reverse T3, also known as rT3 or 3,3′,5′-Triiodo-L-thyronine, is a biologically inactive form of the thyroid hormone T3 and is produced in the body particularly during periods of stress. Under certain conditions such as fibromyalgia, liver disease, and chronic fatigue syndrome (CFS), more rT3 is produced which minimizes the desirable conversion of thyroxine (T4) to T3. Reverse T3 serum concentrations are measured by LC-MS/MS to monitor impact of these and other diseases on thyroid hormone levels.

Application


  • Reverse T3 solution for thyroid research: Reverse T3 (3,3′,5′-Triiodo-L-thyronine) is utilized in biochemical assays to assess thyroid function and metabolism. Its role as an inactive metabolite of thyroid hormone makes it critical in studies investigating thyroid hormone balance and its impact on metabolic rate (Filetti et al., 1980).

  • High-purity rT3 thyroid hormone analog: Due to its high purity and stability, Reverse T3 is essential in thyroid metabolism research, providing a reliable standard for comparing thyroid activity and understanding its biological effects in health and disease states.

  • Thyroid metabolism research using Reverse T3: rT3 is instrumental in the study of thyroid hormone metabolism pathways, particularly in understanding the conversion processes of T4 to T3 and rT3, aiding in deciphering the complex regulatory mechanisms of thyroid hormones.

  • Reverse T3 in comparative endocrinology studies: The application of Reverse T3 solution extends to comparative endocrinology, where it is used to study the variations and similarities in thyroid hormone metabolism among different species, contributing to broader biological and evolutionary insights.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Skórkowska-Telichowska et al.
Endokrynologia Polska, 63(4), 294-299 (2012-08-31)
Inhabitants living in areas with endemic dietary iodine intake deficiency develop nodular goitre. The aim of our study was to evaluate thyroid morphology and function among adults residing in Lower Silesia and to assess the effect on the thyroid gland
S Ruhla et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 43(2), 130-134 (2010-11-26)
Thyroid dysfunction has been shown to be associated with insulin resistance (IR). This may involve peripheral thyroid hormone metabolism, which is assumed to be reflected by the ratio triiodothyronine/reverse triiodothyronine (T3/rT3-ratio). To explore a potential association between the T3/rT3-ratio and
Ashley Hinther et al.
Environmental science & technology, 44(21), 8314-8321 (2010-10-12)
Nanometals are manufactured to particle sizes with diameters in the nanometer range and are included in a variety of consumer and health products. There is a lack of information regarding potential effects of these materials on aquatic organisms. Amphibians are
Pranati Sar et al.
PloS one, 6(6), e20861-e20861 (2011-06-21)
Thyroid hormones regulate cell proliferation, differentiation as well as apoptosis. However molecular mechanism underlying apoptosis as a result of thyroid hormone signaling is poorly understood. The antiapoptotic role of Senescence Marker Protein-30 (SMP30) has been characterized in response to varieties
Annemieke J Lem et al.
The Journal of clinical endocrinology and metabolism, 97(9), 3170-3178 (2012-06-28)
Age-appropriate reference ranges for thyroid hormones are required for detecting pediatric thyroid dysfunction. Data on thyroid hormones and peripheral thyroid metabolism in short children born small for gestational age (SGA) before and during GH treatment are lacking. Our objectives were

Protocols

Determination of the thyroid hormones T4, T3 and rT3 from plasma using an Ascentis Express RP-Amide cartridge to trap the analytes, and a Phenyl-Hexyl column to resolve them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service