Skip to Content
Merck
All Photos(1)

Documents

S-024

Supelco

HU-210 solution

100 μg/mL in methanol, (Spice Cannabinoid), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H38O3
CAS Number:
Molecular Weight:
386.57
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCCCCCC(C)(C)c1cc(O)c2[C@@H]3CC(CO)=CC[C@H]3C(C)(C)Oc2c1

InChI

1S/C25H38O3/c1-6-7-8-9-12-24(2,3)18-14-21(27)23-19-13-17(16-26)10-11-20(19)25(4,5)28-22(23)15-18/h10,14-15,19-20,26-27H,6-9,11-13,16H2,1-5H3/t19-,20-/m1/s1

InChI key

SSQJFGMEZBFMNV-WOJBJXKFSA-N

General description

HU-210 is a synthetic cannabinoid found in the herbal mixture “Spice.” Illicit use of this Spice cannabinoid includes smoking the herbal product to obtain a psychoactive effect. Structurally similar to (-)-Δ8-THC and 11-Hydroxy-Δ9-THC, HU-210 is considerably more potent than natural THC from cannabis and has an extended duration of action.
In July, 2009, the US Drug Enforcement Agency (DEA) placed HU-210 on its Drugs and Chemicals of Concern list. Since HU-210′s placement on this list, DEA has reported seizures of Spice herbal mixtures in Ohio and Florida. HU-210 is currently listed as a Schedule I controlled substance. In recognition of this regulatory challenge, Cerilliant supplies its certified HU-210 reference standard in a convenient, quantitative, DEA-exempt solution.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eun Sook Chung et al.
Brain research, 1451, 110-116 (2012-03-23)
We investigated the effects of synthetic cannabinoids, WIN55,212-2 and HU210, on LPS-injected rat substantia nigra in vivo. Intranigral injection of LPS resulted in a significant loss of nigral dopaminergic (DA) neurons, as determined by Nissl staining and TH immunohistochemistry. LPS-induced
Adrianne R Wilson-Poe et al.
Pharmacology, biochemistry, and behavior, 103(3), 444-449 (2012-10-16)
Co-administration of opioids and cannabinoids can enhance pain relief even when administered on different days. Repeated systemic administration of morphine has been shown to enhance the antinociceptive effect of tetrahydrocannabinol (THC) administered 12h later, and repeated microinjection of the cannabinoid
L N Maslov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 75(12), 15-18 (2012-01-01)
It was shown that perfusion of the isolated heart of rat with solution containing the CB1- and CB2-receptor agonist HU-210 at concentrations of 0.1 or 1.0 microM/L for a duration of 10 min at 20 min before global ischemia (45
Valentina Chiodi et al.
Neurobiology of disease, 45(3), 983-991 (2011-12-31)
Cannabinoid CB1 receptors (CB1Rs) are known to be downregulated in patients and in animal models of Huntington's disease (HD). However, the functional meaning of this reduction, if any, is still unclear. Here, the effects of the cannabinoid receptor agonist WIN
Ming-hua Cao et al.
PloS one, 7(12), e52921-e52921 (2013-01-04)
Acute pancreatitis (AP), especially severe acute pancreatitis often causes extra-pancreatic complications, such as acute gastrointestinal mucosal lesion (AGML) which is accompanied by a considerably high mortality, yet the pathogenesis of AP-induced AGML is still not fully understood. In this report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service