Skip to Content
Merck
All Photos(1)

Key Documents

P-077

Supelco

Propofol-D17 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D17HO
CAS Number:
Molecular Weight:
195.38
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=C(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])=C([2H])C([2H])=C1C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3/i1D3,2D3,3D3,4D3,5D,6D,7D,8D,9D

InChI key

OLBCVFGFOZPWHH-ACWNEFEHSA-N

General description

Propofol, also known as Diprivan®, is a short-acting, hypnotic drug intravenously administered for the induction and maintenance of general anesthesia and sedation. This internal standard is suitable for a variety of uses including forensic and forensic toxicology analysis, isotope dilution methods, calibrator preparation, and drug monitoring in plasma or whole blood.

Application


  • Propofol-D₁₇ in forensic analysis: Propofol-D₁₇ was utilized to confirm testimonial evidence in serial homicide cases through drug detection in decomposed cadavers, demonstrating its value in complex forensic scenarios (Ottaviano et al., 2021).

  • Pharmacokinetic studies with Propofol-D₁₇: A sensitive LC-APCI-MS/MS method was developed using Propofol-D₁₇ to quantify propofol in a bioequivalence study among healthy Chinese subjects, enhancing understanding of its pharmacokinetics (Tang et al., 2020).

  • Evaluation of analytical methods using Propofol-D₁₇: Comparative analysis of LC-MS/MS and GC-MS for propofol detection highlights Propofol-D₁₇′s role in improving analytical procedures for forensic and clinical toxicology (Vaiano et al., 2015).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Diprivan is a registered trademark of Abraxis Bioscience, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranjith Babu et al.
Journal of neuro-oncology, 119(1), 177-185 (2014-05-20)
Adult malignant brainstem gliomas (BSGs) are poorly characterized due to their relative rarity. We have examined histopathologically confirmed cases of adult malignant BSGs to better characterize the patient and tumor features and outcomes, including the natural history, presentation, imaging, molecular
Mirko Nitsche et al.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al], 190(8), 715-721 (2014-05-20)
Recent studies have demonstrated low regional recurrence rates in early-stage breast cancer omitting axillary lymph node dissection (ALND) in patients who have positive nodes in sentinel lymph node dissection (SLND). This finding has triggered an active discussion about the effect
Rosane Menezes Faria Dutra et al.
Clinics (Sao Paulo, Brazil), 69(5), 314-318 (2014-05-20)
In this observational study, we evaluated the peripheral oxygen saturation (SpO2), heart rate, and blood pressure of children with cyanotic congenital heart disease who were undergoing dental extraction. Forty-four patients between the ages of 6 and 12 years who underwent
Joseph R Bardeen et al.
Journal of behavior therapy and experimental psychiatry, 45(3), 415-420 (2014-05-20)
Emotional avoidance has been found to be associated with higher levels of anxiety. However, no research to date has differentiated between the avoidance of positive and negative emotions in relation to anxiety. Additionally, no studies have examined the extent to
Min Dong et al.
British journal of clinical pharmacology, 78(5), 1102-1112 (2014-05-20)
The purpose of this study was to develop a population pharmacokinetic and pharmacodynamic (PK-PD) model for mycophenolic acid (MPA) in paediatric renal transplant recipients in the early post-transplant period. A total of 214 MPA plasma concentrations-time data points from 24

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service