Skip to Content
Merck
All Photos(1)

Key Documents

P-011

Supelco

(+)-Propoxyphene solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO2
CAS Number:
Molecular Weight:
339.47
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@](Cc1ccccc1)([C@H](C)CN(C)C)c2ccccc2

InChI

1S/C22H29NO2/c1-5-21(24)25-22(18(2)17-23(3)4,20-14-10-7-11-15-20)16-19-12-8-6-9-13-19/h6-15,18H,5,16-17H2,1-4H3/t18-,22+/m1/s1

InChI key

XLMALTXPSGQGBX-GCJKJVERSA-N

Gene Information

human ... OPRM1(4988)

General description

A certified Snap-N-Spike® solution standard suitable for propoxyphene testing methods by GC/MS or LC/MS in pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis. Propoxyphene, also known as dextropropoxyphene, is an opioid analgesic drug intended for treatment of mild pain. In combination with either paracetamol or aspirin, dextropropoxyphene is sold under trade names including Darvocet-N® and Darvon®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Darvocet-N is a registered trademark of AAIPharma LLC
Darvon is a registered trademark of Xanodyne Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith Hawton et al.
PLoS medicine, 9(5), e1001213-e1001213 (2012-05-17)
The analgesic co-proxamol (paracetamol/dextropropoxyphene combination) has been widely involved in fatal poisoning. Concerns about its safety/effectiveness profile and widespread use for suicidal poisoning prompted its withdrawal in the UK in 2005, with partial withdrawal between 2005 and 2007, and full
Stephen Winbery et al.
Southern medical journal, 104(7), 533-539 (2011-09-03)
Medicare Part D data from the Quality Improvement Organization's 9th Statement of Work drug safety indicator project under the direction of the Centers for Medicare & Medicaid Services define the potentially inappropriate medications (PIMs) list for Tennessee. These data reveal
Surendra Kumar Mattoo et al.
General hospital psychiatry, 35(1), 100-101 (2012-12-04)
As an infrequent symptom diplopia has been reported with opiate withdrawal, especially heroin, but not dextropropoxyphene. We report incomitant esotropia and diplopia in a case with dextropropoxyphene withdrawal that resolved completely with the resolution of the opiate withdrawal syndrome.
J Bedson et al.
European journal of pain (London, England), 17(3), 434-443 (2012-08-07)
Numerous national guidelines have been issued to assist general practitioners' safe analgesic prescribing. Their effectiveness is unclear. The objective of this study was to examine trends in general practitioners' prescribing behaviour in relation to national guidelines. This was a retrospective
Margareeta Häkkinen et al.
Forensic science international, 222(1-3), 327-331 (2012-08-14)
There is a rising trend of fatal poisonings due to medicinal opioids in several countries. The present study evaluates the drug and alcohol findings as well as the cause and manner of death in opioid-related post-mortem cases in Finland from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service