Skip to Content
Merck
All Photos(1)

Key Documents

M-030

Supelco

Morphine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35

InChI

1S/C17H19NO3/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18/h2-5,10-11,13,16,19-20H,6-8H2,1H3/t10-,11+,13-,16-,17-/m0/s1

InChI key

BQJCRHHNABKAKU-KBQPJGBKSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

Morphine is a very potent opiate analgesic drug used to relieve both acute and chronic severe pain. Sold under trade names including MS Contin®, Kadian®, Oramorph®, and Roxanol, morphine is also a commonly abused recreational drug due to its widespread availability and high potential for addiction. This certified analytical reference standard is suitable for a variety of GC/MS or LC/MS applications from pain prescription monitoring, clinical toxicology and urine drug testing to pharmaceutical research and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Kadian is a registered trademark of Actavis Elizabeth LLC
MS Contin is a registered trademark of Purdue Frederick Company
Oramorph is a registered trademark of AAIPharma, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thitima Chinachoti et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 96(5), 589-594 (2013-06-12)
Presently, in Siriraj Hospital, intrathecal morphine is routinely used in spinal anesthesia for postoperative pain control in more than 600 cases per month with doses lower than 0.3 mg. However, the incidence of side effects is high. This retrospective data
A Bilir
Clinical and experimental obstetrics & gynecology, 40(1), 118-121 (2013-06-04)
Pain management has a particular importance after Cesarean section. This study was undertaken in order to document the efficacy and side-effects of epidural morphine instead of patient-controlled analgesia technique used for the control of post-cesarean pain during postoperative 24-48 hours.
In response.
Alain Gorgeat
Anesthesia and analgesia, 116(6), 1391-1391 (2013-07-12)
Mean versus median in tables and figures.
Tim Thomas Joseph et al.
Anesthesia and analgesia, 116(6), 1390-1391 (2013-05-28)
Wirzafeldi Sawi et al.
Middle East journal of anaesthesiology, 22(1), 21-26 (2013-07-10)
This was a double-blinded, prospective randomized controlled trial to compare the postoperative analgesia, side effects profile and overall satisfaction in patients who received intrathecal fentanyl with or without morphine for elective Caesarean. Sixty ASA I and II parturients were randomized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service