Skip to Content
Merck
All Photos(1)

Documents

L-907

Supelco

Lormetazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Cl2N2O2
CAS Number:
Molecular Weight:
335.18
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN1C(=O)C(O)N=C(c2ccccc2Cl)c3cc(Cl)ccc13

InChI

1S/C16H12Cl2N2O2/c1-20-13-7-6-9(17)8-11(13)14(19-15(21)16(20)22)10-4-2-3-5-12(10)18/h2-8,15,21H,1H3

InChI key

FJIKWRGCXUCUIG-UHFFFAOYSA-N

General description

Lormetazepam is a benzodiazepine with sedative-hypnotic, anxiolytic, anticonvulsant, and muscle relaxant effects. This certified solution standard is suitable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis. Lormetazepam is sold under trade names Loramet and Noctamid for treatment of insomnia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hervé Allain et al.
European journal of clinical pharmacology, 59(3), 179-188 (2003-05-21)
In elderly patients, both falls and impaired memory are considerable medical problems. Hypnotics, which are frequently administered to this patient group for the treatment of insomnia, should ideally not impair equilibrium or memory functions. This double-blind, randomised, four-way, cross-over study
Musical hallucinations during a treatment with benzodiazepine.
François Curtin et al.
Canadian journal of psychiatry. Revue canadienne de psychiatrie, 47(8), 789-790 (2002-11-08)
[Paroxetine-induced severe hyponatremic rhabdomyolisis].
M Portilla-Botelho et al.
Medicina intensiva, 32(7), 365-366 (2008-10-10)
Luc Staner et al.
Psychopharmacology, 181(4), 790-798 (2005-07-19)
Most studies that investigated the next-day residual effects of hypnotic drugs on daytime driving performances were performed on healthy subjects and after a single drug administration. In the present study, we further examine whether the results of these studies could
W Dimpfel et al.
European journal of medical research, 3(10), 453-460 (1998-10-01)
Sleep in humans is classically assessed by recording a multichannel electroencephalogram (EEG) in connection with an electrooculogram (EOG) and an electromyogram (EMG). In general, human sleep is manually staged into 6 categories (from awake through REM sleep to stage 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service