This product is a deuterated form of 7-Hydroxymitragynine prepared in methanol. It is intended for use as an analytical standard and would not be suitable or approved for in vitro or in vivo use. A physiologic dosage would not be relevant or established for this type of product. For analytical purposes, the minimal sample volume for quantitative applications is 1 uL. For more information regarding this product please view the Certificate of Analysis: https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/cer/H-109_FN05252124.pdf.
H-109
7-Hydroxymitragynine-D3 solution
100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
Deuterated 7-Hydroxymitragynine Standard
About This Item
Recommended Products
grade
certified reference material
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol with 0.1N NH3
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−70°C
SMILES string
O[C@@]12C3=C(OC([2H])([2H])[2H])C=CC=C3N=C1[C@@]4([H])N(C[C@@H](CC)[C@@H](/C(C(OC)=O)=C\OC)C4)CC2
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Guten Tag Wie ist denn die Dosierung von diesem Extrakt ? Hier steht leider nichts zur Anwendung Und auch so keine erfahrungs Berichte
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service