Skip to Content
Merck
All Photos(1)

Documents

H-073

Supelco

25-Hydroxyvitamin D2 solution

50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

25-Hydroxyvitamin D2 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O2
CAS Number:
Molecular Weight:
412.65
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

50 μg/mL in ethanol

application(s)

clinical testing
clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

CCCC(O)CC.CC(\C=C\C(C)C(C)(C)O)C1CCC2C(CCCC12C)\C=C\C=C/C=C

InChI

1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24+,25-,26+,28-/m1/s1

InChI key

KJKIIUAXZGLUND-ICCVIKJNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® of 25-Hydroxyvitamin D2, a prohormone whose blood serum levels are used to determine Vitamin D nutritional status. This solution standard is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ.

The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike® format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Application

25-Hydroxyvitamin D2 may be used as a certified reference material (CRM) for the determination of the analyte in biological samples by chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin D Cashman et al.
The British journal of nutrition, 112(2), 193-202 (2014-05-02)
It has been suggested that vitamin D₂ is not very prevalent in the human food chain. However, data from a number of recent intervention studies suggest that the majority of subjects had measurable serum 25-hydroxyvitamin D₂ (25(OH)D₂) concentrations. Serum 25(OH)D₂
Paul Haggarty et al.
The British journal of nutrition, 109(5), 898-905 (2012-11-22)
The aims of the present study were to determine compliance with current advice on vitamin D and to assess the influence of season, dietary intake, supplement use and deprivation on vitamin D status in pregnant mothers and newborns in the
Grant R Campbell et al.
PLoS pathogens, 8(11), e1003017-e1003017 (2012-11-21)
Toll-like receptors (TLR) are important in recognizing microbial pathogens and triggering host innate immune responses, including autophagy, and in the mediation of immune activation during human immunodeficiency virus type-1 (HIV) infection. We report here that TLR8 activation in human macrophages
Stephen J Bruce et al.
Rapid communications in mass spectrometry : RCM, 27(1), 200-206 (2012-12-15)
The aim of the work was to develop and validate a method for the quantification of vitamin D metabolites in serum using ultra-high-pressure liquid chromatography coupled to mass spectrometry (LC/MS), and to validate a high-resolution mass spectrometry (LC/HRMS) approach against
Amin Salehpour et al.
Nutrition journal, 11, 78-78 (2012-09-25)
Vitamin D concentrations are linked to body composition indices, particularly body fat mass. Relationships between hypovitaminosis D and obesity, described by both BMI and waist circumference, have been mentioned. We have investigated the effect of a 12-week vitamin D3 supplementation

Articles

Vitamin D deficiency affects bone health, highlighting its vital role in calcium absorption and bone growth.

Vitamin D deficiency affects bone health, highlighting its vital role in calcium absorption and bone growth.

Vitamin D deficiency affects bone health, highlighting its vital role in calcium absorption and bone growth.

Vitamin D deficiency affects bone health, highlighting its vital role in calcium absorption and bone growth.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service