Skip to Content
Merck
All Photos(1)

Key Documents

F-005

Supelco

Furosemide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

2-8°C

SMILES string

NS(=O)(=O)c1cc(C(O)=O)c(NCc2ccco2)cc1Cl

InChI

1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)

InChI key

ZZUFCTLCJUWOSV-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

General description

Furosemide is a loop diuretic prescribed for treatment of edemas and congestive heart failure. In sports testing settings, furosemide is used as an illicit masking agent in the testing of performance-enhancing drugs. This certified solution standard is suitable for use in LC/MS or GC/MS applications from clinical toxicology, urine drug testing, and forensic analysis to sports testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Carels et al.
JBR-BTR : organe de la Societe royale belge de radiologie (SRBR) = orgaan van de Koninklijke Belgische Vereniging voor Radiologie (KBVR), 96(1), 17-18 (2013-04-25)
We report a case of sudden onset of respiratory distress caused by pulmonary edema due to laryngospasm. The diagnosis was established by the clinical context and chest X-ray. A CT-scan was performed to narrow down the differential diagnosis and to
[What the Furosemide Teva situation did not tell us].
Jean-Yves Nau
Revue medicale suisse, 9(391), 1334-1335 (2013-07-24)
Nida Zaidi et al.
The journal of physical chemistry. B, 117(9), 2595-2604 (2013-02-27)
Exogenous substances like drugs, when absorbed, enter into the circulatory system and bind reversibly and extensively to human serum albumin (HSA). But transport of various drugs like a diuretic, furosemide (FUR), via albumin in uremia is seriously compromised due to
An 82-year-old woman with progressive dyspnea and bilateral infiltrates.
Matthew Koslow et al.
Chest, 143(2), 571-574 (2013-02-06)
Caesarian dissection: a case of chest pain and dyspnea during delivery.
Andreas C Mauer et al.
Circulation, 127(14), 1530-1535 (2013-04-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service