Skip to Content
Merck
All Photos(1)

Key Documents

LM1100

Avanti

12:0-13:0 PE

Avanti Research - A Croda Brand LM1100, methanol solution

Synonym(s):

1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H60NO8P
CAS Number:
Molecular Weight:
593.77
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1100-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1100

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O

InChI key

PGDKJXWZRREZOX-MUUNZHRXSA-N

General description

Phosphatidylethanolamine is the second most abundant phospholipid, essential for various cellular processes. It is a non-bilayer forming phospholipid.

Application

12:0-13:0 PE has been used to study the lipid′s interaction with the membrane protein of interest and as a quantitative lipid standard to monitor various lipid extracts concentration.

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a vital role in oxidative phosphorylation, mitochondrion stability and membrane fusion. It functions as a chaperone by aiding in folding certain membrane proteins. PE contributes to the process of autophagy.

Packaging

2 mL Amber Glass Sealed Ampule (LM1100-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylethanolamine Metabolism in Health and Disease
Calzada E, et al.
Nutrition, 321, 29-88 (2016)
Identifying key membrane protein lipid interactions using mass spectrometry.
Gupta K, et al.
Nature Protocols, 13(5), 1106-1106 (2018)
Akifumi Shiomi et al.
Bioscience, biotechnology, and biochemistry, 84(3), 583-593 (2019-11-26)
Fish cell lines are widely used for the studies of developmental biology, virology, biology of aging, and nutrition physiology. However, little is known about their physicochemical properties. Here, we report the phospholipid compositions and mechanical properties of cell membranes derived
Ethanolamine and Phosphatidylethanolamine Partners in Health and Disease
Patel D and Witt SN
Oxidative Medicine and Cellular Longevity, 2017, 29-88 (2017)
A comprehensive method for lipid profiling by liquid chromatography-ion cyclotron resonance mass spectrometry
Fauland A, et al.
Journal of Lipid Research, 52(12), 2314-2322 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service