Skip to Content
Merck
All Photos(1)

Key Documents

860514P

Avanti

C14 Ceramide (d18:1/14:0)

Avanti Research - A Croda Brand 860514P, powder

Synonym(s):

N-myristoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H63NO3
CAS Number:
Molecular Weight:
509.85
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (860514P-25mg)
pkg of 1 × 5 mg (860514P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860514P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C32H63NO3/c1-3-5-7-9-11-13-15-16-18-19-21-23-25-27-31(35)30(29-34)33-32(36)28-26-24-22-20-17-14-12-10-8-6-4-2/h25,27,30-31,34-35H,3-24,26,28-29H2,1-2H3,(H,33,36)/b27-25+/t30-,31+/m0/s1

InChI key

ZKRPGPZHULJLKJ-JHRQRACZSA-N

General description

Ceramide synthesis is majorly driven by de novo pathway involving the palmitoyl-CoA and L-serine, and also by salvage pathway via recycling of complex sphingolipids. The ceramide transporter, Cert mediates the transfer of C14 from smooth endoplasmic reticulum to Golgi. Ceramide synthase CerS6 enzyme is involved in the synthesis of C14 ceramide.

Application

C14 Ceramide (d18:1/14:0) is suitable for use
  • as a standard in high-performance liquid chromatography/tandem mass spectrometry (LC/MS2) for ceramide quantification from muscel biopsies
  • in lipid monolayer preparation for biophysical studies
  • in the morphological and dimensional evaluation studies

Biochem/physiol Actions

Ceramides regulate biological processes including proliferation, differentiation and apoptosis. In the cystic fibrosis patients, elevated levels of C14 ceramide is observed. It is also a potential biomarker for hepatic steatosis (HS) and C14 ceramide levels correlate with the aspartate aminotransferase (AST) / alanine transaminase (ALT) ratio. Tumor necrosis factor α (TNFα) mediates increase in the C14 levels. Fumonisin B1 prevents the C14 accumulation.

Packaging

5 mL Amber Glass Screw Cap Vial (860514P-25mg)
5 mL Amber Glass Screw Cap Vial (860514P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide effects in the bicelle structure
Barbosa-Barros L, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 317(1-3), 576-584 (2008)
Biochemistry of very-long-chain and long-chain ceramides in cystic fibrosis and other diseases: The importance of side chain
Garic D, et al.
Progress in Lipid Research (2019)
Ceramide N-acyl chain length: a determinant of bidimensional transitions, condensed domain morphology, and interfacial thickness
Dupuy F, et al.
Langmuir, 27(7), 3783-3791 (2011)
Tumor necrosis factor-alpha(TNFalpha)-induced ceramide generation via ceramide synthases regulates loss of focal adhesion kinase (FAK) and programmed cell death
Hernandez-Corbacho M, et al.
The Journal of Biological Chemistry, 290(42), 25356-25373 (2015)
Association between Plasmatic Ceramides Profile and AST/ALT Ratio: C14: 0 Ceramide as Predictor of Hepatic Steatosis in Adolescents Independently of Obesity
Maldonado-Hernandez J, et al.
Canadian journal of gastroenterology & hepatology, 2017 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service