Skip to Content
Merck
All Photos(1)

Key Documents

860066P

Avanti

Brain CPE

Avanti Research - A Croda Brand, powder

Synonym(s):

Ceramide phosphoethanolamine (brain, porcine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H77N2O6P
CAS Number:
Molecular Weight:
689.00
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 25 mg (860066P-25mg)
pkg of 1 × 5 mg (860066P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)COP([O-])(OCC[NH3+])=O

InChI

1S/C38H77N2O6P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-38(42)40-36(35-46-47(43,44)45-34-33-39)37(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,36-37,41H,3-28,30,32-35,39H2,1-2H3,(H,40,42)(H,43,44)/b31-29+/t36-,37+/m0/s1

InChI key

ODXAYQMCAAQOSY-YPDYIYJKSA-N

General description

Ceramide phosphoethanolamine (CPE) is a sphingolipid found in mammalian cells, bacteria and invertebrates. It is a major sphingolipid found in the cell membrane of Drosophila and Musca. A high level of CPE is observed in mammalian brain and testis and lower levels in liver and heart. CPE is a precursor for the production of sphingomyelin in the liver.[1]

Packaging

5 mL Clear Glass Sealed Ampule (860066P-25mg)
5 mL Clear Glass Sealed Ampule (860066P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide phosphoethanolamine, an enigmatic cellular membrane sphingolipid
Panevska A, et al.
Biochimica et Biophysica Acta - Biomembranes (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service