Skip to Content
Merck
All Photos(2)

Key Documents

850476P

Avanti

18:1-18:0 PC

1-oleoyl-2-stearoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1--(9Z-octadecenoyl)-2-octadecanoyl-sn-glycero-3-phosphocholine; OSPC; PC(18:1(9Z)/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H86NO8P
CAS Number:
Molecular Weight:
788.13
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850476P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850476P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O

InChI

1S/C44H86NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20,22,42H,6-19,21,23-41H2,1-5H3/b22-20-/t42-/m1/s1

InChI key

NMJCSTNQFYPVOR-VHONOUADSA-N

General description

18:1-18:0 PC (1-oleoyl-2-stearoyl-sn-glycero-3-phosphocholine) is an asymmetric phospholipid.[1]

Application

18:1-18:0 PC (1-oleoyl-2-stearoyl-sn-glycero-3-phosphocholine) may be used:
  • as a lipid standard in ultra-performance liquid chromatography (UPLC) analyses[2]
  • in bilayer membranes to study the bilayer phase transitions[1]
  • in liposome as substrate to study the transacylase activities of different lysosomal phospholipase A2 (LPLA2) variants[3]

Biochem/physiol Actions

Phosphatidylcholine (PC) intake through diet can enhance the learning and memory ability in rats.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850476P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service