Skip to Content
Merck
All Photos(1)

Key Documents

810384P

Avanti

TopFluor TMR PI(4,5)P2

Avanti Research - A Croda Brand 810384P, powder

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3a,4a-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3-phosphoinositol-4.5-bisphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H92BF2N6O21P3
CAS Number:
Molecular Weight:
1303.07
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (with stopper and crimp cap (810384P-50ug))
pkg of 2 × 50 μg (with stopper and crimp cap (810384P-100ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810384P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylinositol 4,5-bisphosphate/(PI(4,5)P2 or PIP2) is one of the major constituent of the inner leaflet of the plasma membrane, especially in eukaryotic cells. It is an important phosphoinositide phosphate species in mammalian cells.[1]

Biochem/physiol Actions

Phosphatidylinositol 4,5-bisphosphate/(PI(4,5)P2 or PIP2) actively participates in numerous cellular signaling pathways. It acts as a precursor of phosphatidylinositol (3,4,5)-trisphosphate (PI(3,4,5)P3). It helps in the assembly of several viruses, like human immunodeficiency virus 1 (HIV1).[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810384P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (810384P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multivalent cation-bridged PI (4, 5) P2 clusters form at very low concentrations
Wen Y, et al.
Biophysical Journal, 114(11), 2630-2639 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service