Skip to Content
Merck
All Photos(1)

Key Documents

810227P

Avanti

C12-NBD Lactosyl Ceramide

Avanti Research - A Croda Brand 810227P, powder

Synonym(s):

N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-D-lactosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H81N5O16
CAS Number:
Molecular Weight:
984.18
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (810227P-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810227P

shipped in

dry ice

storage temp.

−20°C

General description

C12-NBD Lactosyl ceramide is a fluorescent analog of biologically available compound lactosyl ceramide. Lactosyl ceramideis generally present on neutrophils and macrophages.

Biochem/physiol Actions

Lactosyl ceramide is the major precursor for synthesis of various glycosphingolipids like oligoglycosylceramides and gangliosides. It is an important signaling molecule that is involved in adhesion, migration, cell proliferation and angiogenesis. Lactosyl ceramide is vital for osteoclastogenesis mediated by macrophage-colony stimulating factor.

Packaging

5 mL Amber Glass Screw Cap Vial (810227P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iwamoto et al.
The Journal of biological chemistry, 276(49), 46031-46038 (2001-10-11)
Glycosphingolipids and their metabolites play important roles in a variety of biological processes. Several signal molecules are localized in a glycolipid-enriched microdomain on the cell surface, and their signals are regulated by the glycolipid composition. However, the function of glycolipids
S Hakomori et al.
Journal of biochemistry, 118(6), 1091-1103 (1995-12-01)
Glycosphingolipids (GSLs), cell type-specific markers which change dramatically during ontogenesis and oncogenesis, have been implicated as playing major roles in cellular interactions and control of cell proliferation in multicellular organisms. These functional roles have been partially clarified through two types
W I Weis et al.
Annual review of biochemistry, 65, 441-473 (1996-01-01)
Lectins are responsible for cell surface sugar recognition in bacteria, animals, and plants. Examples include bacterial toxins; animal receptors that mediate cell-cell interactions, uptake of glycoconjugates, and pathogen neutralization; and plant toxins and mitogens. The structural basis for selective sugar
F Pincet et al.
Biophysical journal, 80(3), 1354-1358 (2001-02-27)
Carbohydrate-carbohydrate interactions are rarely considered in biologically relevant situations such as cell recognition and adhesion. One Ca(2+)-mediated homotypic interaction between two Lewis(x) determinants (Le(x)) has been proposed to drive cell adhesion in murine embryogenesis. Here, we confirm the existence of
Subroto Chatterjee et al.
Biochimica et biophysica acta, 1780(3), 370-382 (2007-12-14)
Although lactosylceramide (LacCer) plays a pivotal role in the biosynthesis of nearly all the major glycosphingolipids, its function in regulating cellular function has begun to emerge only recently. Our current opinion is that several physiologically critical molecules such as modified/oxidized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service