Skip to Content
Merck
All Photos(2)

Key Documents

810165P

Avanti

18:1-06:0 NBD PG

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62N5O13P
CAS Number:
Molecular Weight:
803.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810165P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylglycerol (PG) is an anionic phospholipid, constituting 20−25% in E. coli membranes. Nitrobenzoxadiazole phosphoglycerol (NBD PG) is a fluorescently labeled phospholipid with 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) attached to the fatty acid chain of the phosphatidylglycerol.

Application

18:1-06:0 NBD PG may be used in the determination of phosphatidylglycerol distribution across the bilayer of the membrane and to measure the rate constant for transbilayer movement of lipid molecules by preparing giant unilamellar vesicles (GUVs).

Biochem/physiol Actions

Phosphatidylglycerol is involved in the biosynthesis of cardiolipin. It also contributes to the photosynthetic activity by maintaining the structural photosynthetic apparatus.

Packaging

5 mL Amber Glass Screw Cap Vial (810165P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Plant & Cell Physiology null
The role of membrane tension in the action of antimicrobial peptides and cell-penetrating peptides in biomembranes
Hasan M, et al.
Biophysical Reviews null
Specificity of the transport of lipid II by FtsW in Escherichia coli
Mohammadi T, et al.
Journal of biological and chemical chronicles null
Effect of membrane tension on transbilayer movement of lipids
Hasan M, et al.
J. Chem. Phys. null
Direct Evidence for Requirement of Phosphatidylglycerol in Photosystem II of Photosynthesis
Hagio M, et al.
Plant Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service