Skip to Content
Merck
All Photos(1)

Key Documents

800725P

Avanti

N-P Tyrosine PA

Avanti Research - A Croda Brand 800725P, powder

Synonym(s):

N-palmitoyl-tyrosine phosphoric acid (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48N3O7P
CAS Number:
Molecular Weight:
533.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (800725P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800725P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidic acid (LPA) receptor modulators include N-palmitoyl serine phosphoric acid and N-palmitoyl-tyrosine phosphoric acid. N-palmitoyl serine phosphoric acid and N-palmitoyl-tyrosine phosphoric acid are competitive inhibitors of the LPA receptor in Xenopus oocytes. However, in mammalian cells, N-palmitoyl-tyrosine phosphoric acid may act as an agonist for the LPA receptor. LPA is a lipid mediator that acts similar to growth factors through G-protein coupled plasma membrane receptors. LPA may play a role in platelet aggregation, smooth muscle contraction, vasoactive changes, cytoskeletal reorganization and cell proliferation.

Packaging

5 mL Amber Glass Screw Cap Vial (800725P-1mg)

Preparation Note

Product use: N-palmitoyl-serine and N-palmitoyl-tyrosine phosphoric acid can be used for cell studies. dissolved these lipids in 0.1 mL PBS containing 0.1 mg/mL human serum albumin before adding to cells. In X. laevis studies, these LPA inhibitors were dissolved in DMSO at 1 mM and filtered through a 0.45 mM membrane filter before injection.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis of phosphatidylserine-exposing red blood cells by secretory phospholipase A2 generates lysophosphatidic acid and results in vascular dysfunction.
Neidlinger, N.A, et. al.
The Journal of Biological Chemistry, 281, 775-781 (2006)
Inhibitors of lipid phosphatidate receptors: N-palmitoyl-serine and N-palmitoyl-tyrosine phosphoric acids
Bittman, R, et. al.
Journal of Lipid Research, 37, 391-398 (1996)
Recombinant human G protein-coupled lysophosphatidic acid receptors mediate intracellular calcium mobilization
An, S
Molecular Pharmacology, 54, 881-888 (1998)
R Bittman et al.
Journal of lipid research, 37(2), 391-398 (1996-02-01)
An improved synthesis of two lipid phosphoric acids, N-palmitoyl-L-serine phosphoric acid (NP-Ser-PA) and N-palmitoyl-L-tyrosine phosphoric acid (NP-Tyr-PA), from the benzyl esters of L-serine and L-tyrosine is described. The sequence of N-acylation, followed by phosphitylation with N, N-diisopropyl dibenzyl phosphoramidite, oxidation
Hydrolysis of phosphatidylserine-exposing red blood cells by secretory phospholipase A2 generates lysophosphatidic acid and results in vascular dysfunction
Neidlinger, N.A
The Journal of Biological Chemistry, 281, 775-781 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service