Skip to Content
Merck
All Photos(2)

Key Documents

700107P

Avanti

Sitosterol (synthetic from plant sterol)

Avanti Research - A Croda Brand

Synonym(s):

Synthetic sterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
UNSPSC Code:
12352211
NACRES:
NA.28

Assay

>99% (TLC)

form

powder

packaging

pkg of 1x1 g (700107P-1G)
pkg of 1x100 mg (700107P-100MG)
pkg of 1x200 mg (700107P-200MG)
pkg of 1x500 mg (700107P-500MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@H](CC[C@@H](CC)C(C)C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4CC(O)CC[C@]4(C)[C@@]3([H])CC[C@]12C

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23?,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-BWRKXDIJSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (700107P-100MG)
5 mL Amber Glass Screw Cap Vial (700107P-200MG)
20 mL Amber Glass Screw Cap Vial (700107P-500MG)
20 mL Amber Glass Screw Cap Vial (700107P-1G)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service