Skip to Content
Merck
All Photos(1)

Key Documents

700066P

Avanti

7-dehydrocholesterol

Δ,5,7-cholesterol, powder

Synonym(s):

7-dehydrocholesterin; Provitamin D3; 110807

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700066P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700066P

shipped in

dry ice

storage temp.

−70°C

SMILES string

OC1CCC2(C3CCC4(C(CCC4C(CCCC(C)C)C)C3=CC=C2C1)C)C

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9-10,18-19,21,23-25,28H,6-8,11-17H2,1-5H3

InChI key

UCTLRSWJYQTBFZ-UHFFFAOYSA-N

General description

7-dehydrocholesterol is a metabolite produced in cholesterol synthesis. It is an unsaturated sterol predominantly found in the epidermis.

Application

7-dehydrocholesterol has been used:
  • in gas chromatography/mass spectrometry (GC/MS)-based sterol profiling to quantify 7-dehydrocholesterol in oligodendrocyte progenitor cells (OPCs)
  • as an internal standard for the analysis of sterols and oxysterols using ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • as an internal standard for lipid measurements by liquid chromatography (LC)-MS

Biochem/physiol Actions

7-dehydrocholesterol acts as a precursor of vitamin D3 under the conditions of high temperature and ultraviolet (UV) light.

Packaging

5 mL Amber Glass Screw Cap Vial (700066P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosynthesis of Vitamin D in Skin
Principles of Medical Biology (1997)
Galya Staneva et al.
Langmuir : the ACS journal of surfaces and colloids, 32(6), 1591-1600 (2016-01-20)
7-Dehydrocholesterol, an immediate metabolic predecessor of cholesterol, can accumulate in tissues due to some metabolic abnormalities, causing an array of symptoms known as Smith-Lemli-Opitz syndrome. Enrichment of cellular membranes with 7-dehydrocholesterol interferes with normal cell-signaling processes, which involve interaction between
Manish Sethi et al.
Nanoscale, 6(4), 2321-2327 (2014-01-15)
The effects of nanoparticle (NP) properties, such as size, shape and surface charge, on their efficacy and toxicity have been studied extensively. However, the effect of controlled drug release on NP efficacy and toxicity has not been thoroughly evaluated in
Diverse Chemical Scaffolds Enhance Oligodendrocyte Formation by Inhibiting CYP51, TM7SF2, or EBP
Allimuthu D, et al.
Cell Chemical Biology, 26(4), 593-599 (2019)
Peng He et al.
Biomacromolecules, 6(5), 2455-2457 (2005-09-13)
These studies show that single-walled carbon nanotubes (SWNTs) can be effectively modified using phospholipids. Using a simple surface modification of SWNTs, followed by deposition of 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine (DCPE) phosphipid, results in stable water-dispersible SWNTs with highly uniform thickness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service