Skip to Content
Merck
All Photos(2)

Key Documents

700033P

Avanti

5β,6β-epoxycholestanol

Avanti Research - A Croda Brand

Synonym(s):

110805

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

cholestanol, 5β,6β-epoxy

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700033P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]([C@](CC[C@H](O)C3)(C)[C@]3(O4)[C@H]4C5)[C@@H]5[C@@H]1CC[C@@H]2[C@H](C)CCCC(C)C

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24-27(29-24)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24-,25-,26-,27-/m1/s1

InChI key

PRYIJAGAEJZDBO-DTLXENBRSA-N

General description

5β,6β-epoxycholestanol (5β,6β-EC) is generated by cholesterol epoxidation and is a diastereoisomer of 5α,6α-epoxycholestanol (5α,6α-EC).

Application

5β,6β-epoxycholestanol may be used as a oxy-mix component in treating human colorectal adenocarcinoma CaCo-2 cells to test its effect on toll-like receptor (TLR) expression, in the preparation of langmuir monolayers for isotherm studies, as an internal standard for gas chromatography-mass spectrometry (GC–MS) chromatograms

Biochem/physiol Actions

5β,6β-epoxycholestanol (5β,6β-EC) is associated with human carotid plaques and is not down-regulated by apolipoprotein A1 (apoA-I) protein. 5,6β-EC along with 5α,6α-epoxycholesterol and other oxysterols are reported to be associated with Alzheimer′s disease. It is catabolized by the enzyme cholesterol-5,6-epoxide hydrolase (ChEH) enzyme to 3β,5α,6β-triol. 5β,6β-EC elicits cytotoxic and inflammatory properties.

Packaging

5 mL PTFE Vial with Screw Cap (700033P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced condensing and ordering effects by 7-ketocholesterol and 5beta, 6beta-epoxycholesterol on DPPC monolayers
Telesford DM, et al.
Langmuir, 31(36), 9859-9869 (2015)
A dietary mixture of oxysterols induces in vitro intestinal inflammation through TLR2/4 activation: The protective effect of cocoa bean shells
Rossin D, et al.
Antioxidants, 8(6), 151-151 (2019)
Gas chromatography-mass spectrometry determination of conjugated linoleic acids and cholesterol oxides and their stability in a model system
Yen TY, et al.
Analytical Biochemistry, 400(1), 130-138 (2010)
Fast LC-MS/MS analysis of free oxysterols derived from reactive oxygen species in human plasma and carotid plaque
Helmschrodt C, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 425, 3-8 (2013)
Cholesterol Acceptors Regulate the Lipidome of Macrophage Foam Cells
Paul A, et al.
International Journal of Molecular Sciences, 20(15), 3784-3784 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service