Z676748
ART® low retention barrier tips
Tip design ART 200, Reload insert
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
volume:
200 μL
sterility:
sterile
feature:
barrier
Reload insert
Tip design ART 200
Reload insert
Tip design ART 200
packaging:
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2069-05-RI
Recommended Products
material
colorless
sterility
sterile
feature
barrier
Reload insert
Tip design ART 200
packaging
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename
Molecular BioProducts 2069-05-RI
technique(s)
PCR: suitable
maximum volume
200 μL
volume
200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service