Z676721
ART® barrier tip
maximum volume 200 μL, Tip design ART 200, Reload insert, low retention: no
Synonym(s):
pipet tips, pipette tips
About This Item
low retention: no
Reload insert
Tip design ART 200
Recommended Products
material
colorless
sterility
sterile
feature
barrier
low retention: no
Reload insert
Tip design ART 200
packaging
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename
Molecular BioProducts 2069-RI
technique(s)
PCR: suitable
maximum volume
200 μL
volume
200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required.
Available as reload inserts consisting of sterile pre-loaded blister tubs of 96 tips with or without hinged racks. The empty hinged racks are also available separately.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service