Z676713
ART® low retention barrier tips
Tip design ART 100E, Hinged rack
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier
endotoxin free
Hinged rack
Tip design ART 100E
packaging
pack of 4800 ea (50 hinged racks of 96 tips)
manufacturer/tradename
Molecular BioProducts 2065E-05-HR
technique(s)
PCR: suitable
maximum volume
100 μL
Looking for similar products? Visit Product Comparison Guide
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service