Key Documents
Z675431
Sterile economy sample spoon
1.25 mL
Synonym(s):
dispensing tool
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Pricing and availability is not currently available.
description
PS
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F36740-00001
General description
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
  • Use to collect specific volumes of dry or liquid materials
  • Remains upright when filled or empty for use as a weighing boat
Choose from one of the most recent versions:
Certificates of Analysis (COA)