Key Documents
Z338389
Whatman® GD/XP syringe filters
Nylon membrane
Synonym(s):
Whatman filter, syringe filter, syringe filters
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
Pricing and availability is not currently available.
material
Nylon membrane
polypropylene housing
sterility
non-sterile
feature
Polypropylene prefilter
packaging
pack of 150 ea
manufacturer/tradename
Whatman 6970-2504
Whatman Article No. 28420561 (US reference)
diam.
25 mm
volume
≤25 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
Suitability
Suitable for filtering trace metal analysis samples and for particulate laden sample preparation.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
Whatman is a registered trademark of Cytiva
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)