Skip to Content
Merck

Z231908

Duran® beakers

capacity 2,000 mL, low form

Synonym(s):

211066301, container, flask, glass beaker, jar, vessle

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.43
UNSPSC Code:
41121803
Capacity:
2,000 mL
Material:
borosilicate glass 3.3
Feature:
graduations
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

borosilicate glass 3.3

feature

graduations

manufacturer/tradename

DURAN®, Schott

availability

not available in USA and Canada

H × O.D.

185 mm × 132 mm

capacity

2,000 mL

General description

With spout. Meets ISO 3819 and DIN 12331 standards.

Features and Benefits

  • Complies with ISO 3819
  • Manufactured from borosilicate glass 3.3
  • Spout for clean pouring
  • Fired-on, highly durable white ceramic
  • Easy-to-read scale with graduation marks to estimate the content

Other Notes

Browse all glass beakers

Legal Information

DURAN is a registered trademark of DWK Life Sciences


Still not finding the right product?

Explore all of our products under Duran® beakers




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library