Key Documents
Z210552
Pro-Pak® distillation packing
0.24 in.2, 316 SS
Synonym(s):
Distillation Packing Material
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.
packaging
pkg of 1 L
manufacturer/tradename
Ace Glass 662423
General description
Efficient, self-wetting, protruded metal dump-packing for distillation columns of 0.75 to 12 in. diameter. Maximum reflux rate of approximately 3.4 L/h. Free space for 0.16 in.2 size is 94% and for 0.24 in.2 size is 96%. Each package has a packing volume of approx. 1 L. Packed heights up to 20 ft possible without distortion.
Legal Information
Pro-Pak is a registered trademark of Cannon Instruments Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)