Key Documents
Z176982
Sigma-Aldrich
Scienceware® serum-vial support
Holds 40 x 16-20 mm tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
Pricing and availability is not currently available.
material
polypropylene
feature
Holds 40 x 16-20 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18513-2040
L × W × H
10 in. × 4 3/8 in. × 3 1/2 in.
General description
Lightweight PP. Corrosion-proof. Fits most water baths for elevated temperature incubations. Top shelf height is 41mm (1⁵⁄₈").
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)