Key Documents
WHA77002110
Whatman® UNIFILTER® 384 well filter plates
clear polystyrene, non-sterile, DNA, 50 ea
Synonym(s):
Z746460, micro plate, multiwell plate, well plates
About This Item
UNSPSC Code:
41104900
NACRES:
NB.22
Pricing and availability is not currently available.
Product Name
Whatman® UNIFILTER® plates 384, 384 well DNA binding plate, working volume 100 μL, filter bottom with long drip director
material
conical bottom natural wells
natural polypropylene
polystyrene filter
sterility
non-sterile
feature
filter bottom with long drip director
packaging
pkg of 50 ea
manufacturer/tradename
Cytiva
Whatman 7700-2110
Whatman Article No. 28420803 (US reference)
maximum volume
100 μL
size
384 wells
working volume
100 μL
binding type
DNA high binding (membrane)
non-treated surface
General description
Cytiva′s Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Polystyrene microplate for clean-up and cell capture
A high-throughput microplate can facilitate cell capture or dye removal and other sample clean-up processes requiring high recovery. Whatman UNIFILTER microplates are a good choice because of their long drip director and their ability to support gel filtration media.
Features and Benefits
  • Usable with gel filtration media, allowing high-throughput removal of dye blobs
  • Long drip director prevents well-to-well cross contamination
  • Rigid, clear polystyrene microplate can withstand centrifugation
  • Membrane media option of DNA binding polymer or glass microfiber
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)