Key Documents
WHA10418412
Whatman® Nuclepore Track-Etched Membranes
polycarbonate filter, filter diam. 47 mm, pore size 10 μm, non-sterile, case of 100 ea
About This Item
UNSPSC Code:
41104921
NACRES:
NB.24
material
polycarbonate filter
sterility
non-sterile
packaging
case of 100 ea
manufacturer/tradename
Cytiva 10418412
filter diam.
47 mm
pore size
10 μm
Application
  • Epifluorescence microscopy
  • Environmental analysis
  • Cell biologyEPA testing
  • Fuel testing
  • Bioassays
  • Parasitology
  • Air analysis
  • Water microbiology
Legal Information
Nuclepore is a trademark of Cytiva
Whatman is a registered trademark of Cytiva