Skip to Content
Merck
All Photos(1)

Key Documents

W509531

Sigma-Aldrich

Methyl palmitate

≥97%

Synonym(s):

n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2CH3
CAS Number:
Molecular Weight:
270.45
Beilstein:
1780973
EC Number:
Council of Europe no.:
581
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.180
NACRES:
NA.21
biological source:
Cocos nucifera
palm oil
synthetic
food allergen:
tree nuts

biological source

Cocos nucifera
palm oil
synthetic

Quality Level

Assay

≥97%

refractive index

n20/D 1.4512 (lit.)

bp

185 °C/10 mmHg (lit.)

mp

32-35 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

Organoleptic

waxy

SMILES string

CCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3

InChI key

FLIACVVOZYBSBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl palmitate is a fatty acid methyl ester (FAME) that occurs naturally in crude palm oil and walnut.

Application

Methyl palmitate can be used as an additive in food and cosmetic industries for the stabilization of fats and oil products by slowing down the auto-oxidation of unsaturated fatty acids.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent free synthesis of methyl palmitate over sulfated zirconia solid acid catalyst
Saravanan K, et al.
Fuel: The Science and Technology of Fuel and Energy, 165(11), 298-305 (2016)
Density and speed of sound measurements on five fatty acid methyl esters at 83 kPa and temperatures from (278.15 to 338.15) K.
Ott LS, et al.
Journal of Chemical and Engineering Data, 53(10), 2412-2416 (2008)
Enzyme catalyzed synthesis of cosmetic esters and its intensification: A review
Khan NR and Rathod VK
Process. Biochem., 50(11), 1793-1806 (2015)
Biodiesel production from crude Jatropha curcas L. seed oil with a high content of free fatty acids.
Berchmans HJ & Hirata S.
Bioresource Technology, 99(6), 1716-1721 (2008)
Markus Rentsch et al.
Transplant international : official journal of the European Society for Organ Transplantation, 18(9), 1079-1089 (2005-08-17)
Inhibition or destruction of Kupffer cells (KC) may protect against ischemia-reperfusion (IR) induced primary graft nonfunction (PNF) in liver transplantation. Besides KC activation, PNF is characterized by microvascular perfusion failure, intrahepatic leukocyte accumulation, cell death and hepatocellular dysfunction. KCs can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service